Merck
  • The effects of mycotoxins and selenium deficiency on tissue-engineered cartilage.
The effects of mycotoxins and selenium deficiency on tissue-engineered cartilage.
Cells, tissues, organs (2012-04-28)
Minling Lu, Junling Cao, Fuqiang Liu, Siyuan Li, Jinghong Chen, Qiang Fu, Zengtie Zhang, Jiayuan Liu, Mingxiu Luo, Jiali Wang, Jin Li, Bruce Caterson
ABSTRACT
To investigate the effects of 3 mycotoxins, deoxynivalenol (DON), nivalenol (NIV) and T-2 toxin, in the presence and absence of selenium (Se) on the metabolism of tissue-engineered cartilage to mimic conditions found in Kashin-Beck disease (KBD) environments. Chondrocytes were seeded onto bone matrix gelatin (BMG) to construct engineered cartilage. The 3 toxins were added to the culture media for 3 weeks followed by immunhistochemical analyses of collagens type II and X, aggrecan, matrix metalloproteinases 1 and 3 (MMP-1 and MMP-3), MMP inhibitors 1 and 3 (TIMP-1 and TIMP-3) and α(2) macroglobulin (α2M). Type II collagen was decreased while type X collagen was increased in response to DON, NIV and T-2 toxin. Aggrecan was reduced by all 3 mycotoxins. Compared with the control, the 3 toxins decreased the expression of α2M, TIMP-1 and TIMP-3, and increased the expression of MMP-1 and MMP-3. Se could partially inhibit the effects of DON, NIV and T-2 toxins. Under the low Se condition, the 3 mycotoxins produced procatabolic changes in cartilage resulting in the loss of aggrecan and type II collagen and promoted a hypertrophic phenotype of chondrocytes characterized by increasing type-X-collagen expression, enhancing the expression of MMPs, while weakening the TIMPs. Se could partially block the effects mentioned above. These results support the hypothesis that the combination of mycotoxin stress and Se deficiency would be the causative factors for KBD.
Product Description
Supelco
Nivalenol solution, 100 μg/mL in acetonitrile, analytical standard
Nivalenol in acetonitrile, IRMM®, certified reference material