- A synthetic double-stranded RNA, poly I:C, induces a rapid apoptosis of human CD34(+) cells.
A synthetic double-stranded RNA, poly I:C, induces a rapid apoptosis of human CD34(+) cells.
Toll-like receptor 3 (TLR3), retinoic acid-inducible gene I, and melanoma differentiation-associated antigen 5 (RIG-I/MDA-5) helicases are known to sense double-stranded RNA (dsRNA) virus and initiate antiviral responses, such as production of type-I interferons (IFNs). Recognition of dsRNA by TLR3 or RIG-I/MDA-5 is cell-type-dependent and recent studies have shown a direct link between TLRs and hematopoiesis. We hypothesized that viral dsRNA recognized by either TLR3 or RIG-I/MDA-5, affects the growth of human hematopoietic stem/progenitor cells. Here we show that polyinosinic polycytidylic acid (poly I:C)-mediated very rapid apoptosis occurs within 1 hour in CD34(+) cells in a dose-dependent manner. Polyadenylic-polyuridylic acid, another synthetic dsRNA that signals only through TLR3, had no effect. Poly I:C-LMW/LyoVec, a complex between low molecular-weight poly I:C and the transfection reagent LyoVec, which signals only through RIG-I/MDA-5, induces apoptosis of CD34(+) cells. A strong and sustained upregulation of messenger RNA and protein levels of Noxa, a proapoptotic BH3-only protein that can be induced by RIG-I/MDA-5 pathway, is found in CD34(+) cells treated by poly I:C. Although poly I:C upregulates type-I IFNs in CD34(+) cells, neither exogenous IFN-α nor IFN-β induces rapid apoptosis in CD34(+) cells and neutralization or blocking of type-I IFN receptor does not rescue CD34(+) cells, whereas Z-VAD, a pan-caspase inhibitor, rescues the cells from apoptosis. These results suggest that RIG-I/MDA-5, but not TLR3, signaling triggers poly I:C-induced rapid apoptosis of human CD34(+) cells, which will provide an insight into the mechanisms of dsRNA virus-mediated hematopoietic disorders.