Merck
  • Detection of the 1H and 15N NMR resonances of sulfamate groups in aqueous solution: a new tool for heparin and heparan sulfate characterization.
Detection of the 1H and 15N NMR resonances of sulfamate groups in aqueous solution: a new tool for heparin and heparan sulfate characterization.
Analytical chemistry (2011-09-15)
Derek J Langeslay, Szabolcs Beni, Cynthia K Larive
ABSTRACT
Sulfamate (NHSO(3)(-)) groups are critically important structural elements of the glycosaminoglycans heparin and heparan sulfate (HS). Experimental conditions are presented for detection of the sulfamate (1)H NMR resonances in aqueous solution. NMR spectra reported for N-sulfoglucosamine (GlcNS) and the synthetic pentasaccharide drug fondaparinux demonstrate the broad utility of the sulfamate group (1)H chemical shifts to reflect differences in molecular structure. The sulfamate protons also provide an efficient route for detection of (15)N chemical shifts through proton-nitrogen correlations measured with the heteronuclear single quantum coherence (HSQC) experiment. The HSQC spectra of GlcNS, fondaparinux, and the low-molecular weight heparin enoxaparin illustrate the power of the (1)H and (15)N chemical shifts of the sulfamate NH groups for the structural characterization of heparin and HS.
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis
Supelco
Sulfamic acid, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%
Sigma-Aldrich
Sulfamic acid, ≥99.5% (alkalimetric)
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%