Skip to Content
Merck
All Photos(3)

Key Documents

228745

Sigma-Aldrich

Ammonium sulfamate

ACS reagent, ≥98.0%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$55.30
500 G
$120.00

$55.30


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
$55.30
500 G
$120.00

About This Item

Linear Formula:
NH4SO3NH2
CAS Number:
Molecular Weight:
114.12
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38060607
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥98.0%
grade:
ACS reagent
form:
crystalline powder
crystalline solid
crystals or chunks

$55.30


In StockDetails


Request a Bulk Order

grade

ACS reagent

Quality Level

Assay

≥98.0%

form

crystalline powder
crystalline solid
crystals or chunks

impurities

≤0.02% insolubles

ign. residue

≤0.10%

pH

4.5-6.0 (25 °C, 114.1 g/L)

mp

131-135 °C (lit.)

cation traces

heavy metals: ≤5 ppm

SMILES string

N.NS(O)(=O)=O

InChI

1S/H3NO3S.H3N/c1-5(2,3)4;/h(H3,1,2,3,4);1H3

InChI key

GEHMBYLTCISYNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium sulfamate can be used as a catalyst to synthesize arylidene derivatives via microwave-enhanced Knoevenagel condensation[1].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Chemical Behavior of Low Valence Sulfur Compounds. III. Production of Ammonium Sulfamate by the Oxidation of Ammonium Thiosulfate.
Naito K, et al.
Bulletin of the Chemical Society of Japan, 43(5), 1365-1372 (1970)
Ammonium Sulfamate as Substitute for Lead Peroxide in Microdetermination of Carbon and Hydrogen.
Hussey AS, et al.
Analytical Chemistry, 27(2), 280-281 (1955)
Microwave-enhanced Knoevenagel condensation catalyzed by NH2SO3NH4
Liu C, et al.
Molecular Catalysis, 258, 367-370 (2006)
S Aleryani et al.
The Journal of biological chemistry, 273(11), 6041-6045 (1998-04-16)
Incubation of S-nitrosocysteine or S-nitrosoglutathione (5-100 M) in the presence of a generator of superoxide (xanthine/xanthine oxidase) resulted in a time-dependent decomposition of S-nitrosothiols and accumulation of nitrite/nitrate in reaction mixtures. Quantitatively, the amounts of nitrite/nitrate represented >90% of nitrosonium
Synthesis of Ammonium Dinitramide by Nitration of Potassium and Ammonium Sulfamate. The Effect of Sulfamate Conterion on ADN Purity.
Nazeri GH, et al.
Iranian Journal of Chemistry and Chemical Engineering, 27(!) (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service