Merck
  • Probing the carbohydrate recognition domain of E-selectin: the importance of the acid orientation in sLex mimetics.
Probing the carbohydrate recognition domain of E-selectin: the importance of the acid orientation in sLex mimetics.
Bioorganic & medicinal chemistry (2009-12-02)
Alexander Titz, John Patton, Martin Smiesko, Zorana Radic, Oliver Schwardt, John L Magnani, Beat Ernst
ABSTRACT
The selectin-leukocyte interaction is the initial event in the early inflammatory cascade. This interplay proceeds via the terminal tetrasaccharide sialyl Lewis(x) (sLe(x)), present on physiological selectin ligands and E- and P-selectins located on the endothelial surface. Blocking this process is regarded as a promising therapeutic approach for inflammatory diseases where excessive leukocyte efflux is responsible for tissue damage. Selectin antagonists are generally based on sLe(x) as lead structure, containing the essential pharmacophores pre-oriented in the bioactive conformation. In this work, we describe a set of competitive sLe(x) mimetics possessing the carboxylic acid pharmacophore equipped with additional hydrophobic substituents as neuraminic acid (Neu5Ac) replacements. This small library of antagonists derived from Huisgen-1,3-dipolar cycloadditions allows to further probe the carbohydrate recognition domain of E-selectin.
Sigma-Aldrich
DL-Isoserine, 98%