Skip to Content
Merck
All Photos(1)

Documents

286338

Sigma-Aldrich

DL-Isoserine

98%

Synonym(s):

(±)-3-Amino-2-hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
105.09
Beilstein:
1721413
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

235 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NCC(O)C(O)=O

InChI

1S/C3H7NO3/c4-1-2(5)3(6)7/h2,5H,1,4H2,(H,6,7)

InChI key

BMYNFMYTOJXKLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Cz Dobrowolski et al.
Physical chemistry chemical physics : PCCP, 12(36), 10818-10830 (2010-07-10)
The IR low-temperature Ar and Kr matrix spectra of l-isoserine were registered for the first time and interpreted by means of the anharmonic DFT frequencies calculated at the B3LYP/aug-cc-pVTZ and B3LYP/aug-cc-pVDZ levels. 54 l-isoserine conformers were predicted to be stable
Nagarjuna Palyam et al.
The Journal of organic chemistry, 74(11), 4390-4392 (2009-05-02)
We report a new protocol for synthesis of L-1-deoxymannojirimycin, L-1-deoxyidonojirimycin, and the N-isopropyl derivative of the latter compound from the readily available precursors (S)-isoserinal hydrate and 2-tert-butyl-2-methyl-1,3-dioxan-5-one. The key steps include diastereoselective proline-catalyzed syn aldol transformation and a reductive amination/cyclization.
B S Coller et al.
The Journal of biological chemistry, 268(28), 20741-20743 (1993-10-05)
Peptides containing sequences derived from the new NH2 terminus of the seven-transmembrane domain thrombin receptor after thrombin cleavage can activate platelets directly. We recently demonstrated that such peptides are readily cleaved and inactivated by plasma, serum, and endothelial cell-associated aminopeptidase
Thomas Rühl et al.
Chemical communications (Cambridge, England), (15)(15), 1630-1631 (2002-08-13)
The photolytic decomposition of trifunctional carbene generating photoaffinity probes in methanolic solution was studied, a cleavage reaction with butylamine in water, the conjugation with a ligand (moenomycin), and experiments that demonstrate that the fully armed probes interact with penicillin-binding protein
J Du et al.
Nucleosides & nucleotides, 17(1-3), 1-13 (1998-08-26)
Asymmetric synthesis of N-substituted oxazolidinyl nucleosides has been accomplished from L-isoserine, trans- and cis-Oxazolidine intermediates (4 and 5) were stereoselectively constructed from N-protected L-isoserine with a menthoxycarbonyl group by the condensation with benzoyloxy acetaldehyde dimethyl acetal in a ratio of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service