Merck
  • Induction of hydroxyl radical production in Trametes versicolor to degrade recalcitrant chlorinated hydrocarbons.
Induction of hydroxyl radical production in Trametes versicolor to degrade recalcitrant chlorinated hydrocarbons.
Bioresource technology (2009-07-21)
Ernest Marco-Urrea, Elisabet Aranda, Gloria Caminal, Francisco Guillén
ABSTRACT
Extracellular hydroxyl radical ((*)OH) production via quinone redox cycling in Trametes versicolor, grown in a chemically defined medium, was investigated to degrade trichloroethylene (TCE), perchloroethylene (PCE), 1,2,4- and 1,3,5-trichlorobenzene (TCB). The activity of the enzymes catalyzing the quinone redox cycle, quinone reductase and laccase, as well as the rate of (*)OH production, estimated as the formation of thiobarbituric acid reactive substances (TBARS) from 2-deoxyribose, increased rapidly during the first 2-3 days and then remained at relatively constant levels. Under quinone redox cycling conditions, TCE degradation was concomitant to TBARS production and chloride release, reaching a plateau after 6 h of incubation. Similar results were obtained in PCE, 1,2,4- and 1,3,5-TCB time course degradation experiments. The mole balance of chloride release and 1,2,4-TCB and TCE degraded suggests that these chemicals were almost completely dechlorinated. Experiments using [(13)C]-TCE confirmed unequivocal transformation of TCE to (13)CO(2). These results are of particular interest because PCE and 1,3,5-TCB degradation in aerobic conditions has been rarely reported to date in bacterial or fungal systems.
Sigma-Aldrich
1,2,4-Trichlorobenzene, anhydrous, ≥99%
Sigma-Aldrich
1,2,4-Trichlorobenzene, suitable for HPLC, ≥99%
Supelco
1,2,4-Trichlorobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
1,2,4-Trichlorobenzene, ReagentPlus®, ≥99%