Merck
  • Truncation of N- and C-terminal regions of Streptococcus mutans dextranase enhances catalytic activity.
Truncation of N- and C-terminal regions of Streptococcus mutans dextranase enhances catalytic activity.
Applied microbiology and biotechnology (2011-04-12)
Young-Min Kim, Ryoko Shimizu, Hiroyuki Nakai, Haruhide Mori, Masayuki Okuyama, Min-Sun Kang, Zui Fujimoto, Kazumi Funane, Doman Kim, Atsuo Kimura
ABSTRACT
Multiple forms of native and recombinant endo-dextranases (Dexs) of the glycoside hydrolase family (GH) 66 exist. The GH 66 Dex gene from Streptococcus mutans ATCC 25175 (SmDex) was expressed in Escherichia coli. The recombinant full-size (95.4 kDa) SmDex protein was digested to form an 89.8 kDa isoform (SmDex90). The purified SmDex90 was proteolytically degraded to more than seven polypeptides (23-70 kDa) during long storage. The protease-insensitive protein was desirable for the biochemical analysis and utilization of SmDex. GH 66 Dex was predicted to comprise four regions from the N- to C-termini: N-terminal variable region (N-VR), conserved region (CR), glucan-binding site (GBS), and C-terminal variable region (C-VR). Five truncated SmDexs were generated by deleting N-VR, GBS, and/or C-VR. Two truncation-mutant enzymes devoid of C-VR (TM-NCGΔ) or N-VR/C-VR (TM-ΔCGΔ) were catalytically active, thereby indicating that N-VR and C-VR were not essential for the catalytic activity. TM-ΔCGΔ did not accept any further protease-degradation during long storage. TM-NCGΔ and TM-ΔCGΔ enhanced substrate hydrolysis, suggesting that N-VR and C-VR induce hindered substrate binding to the active site.
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 10-25 units/mg solid
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 400-800 units/mg protein
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 100-250 units/mg protein