Skip to Content
Merck
All Photos(2)

Key Documents

D5884

Sigma-Aldrich

Dextranase from Penicillium sp.

lyophilized powder, 10-25 units/mg solid

Synonym(s):

1,6-α-D-Glucan 6-glucanohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

10-25 units/mg solid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Dextranase from Penicillium sp. has been used in a study to assess a new method of synthesizing biopolymeric affinity ligands. Dextranase from Penicillium sp. Has also been used in a study to investigate the cloning and sequencing of a dextranase-encoding cDNA from Penicillium minioluteum.
Dextranase from Sigma has been used for the hydrolysis of carbohydate polymers, during the study of polysaccharide synthesis by Phanerochaete chrysosporium. It has also been used in the synthesis of new enzymatically degradable thermo-responsive nanogels.

Biochem/physiol Actions

An endodextranase that hydrolyzes α-(1,6)-glucosidic linkages in dextran. Dextrans are undesirable compounds synthesized from sucrose by microbial contaminants during sugar production. They increase viscosity of the flow and decrease industrial recovery. Dextranase has been used for hydrolyzing dextran at sugar mills in order to improve efficiency of sugar production.

Quality

Crude

Unit Definition

One unit will liberate 1.0 μmole of isomaltose (measured as maltose) per min at pH 6.0 at 37 °C, using dextran as substrate.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Polysaccharide synthesis by Phanerochaete chrysosporium during degradation of kraft lignin
Leisola M, et al.
Europ. J. Appl. Microbiol. Biotechnol., 15(3), 180-184 (1982)
Synthesis of new enzymatically degradable thermo-responsive nanogels.
Aguirre, G., Ramos, J., & Forcada, J
Soft Matter, 9(1), 261-270 (2013)
B Garcia et al.
FEMS microbiology letters, 143(2-3), 175-183 (1996-10-01)
A cDNA from Penicillium minioluteum HI-4 encoding a dextranase (1,6-alpha-glucan hydrolase, EC 3.2.1.11) was isolated and characterized. cDNA clones corresponding to genes expressed in dextran-induced cultures were identified by differential hybridization. Southern hybridization and restriction mapping analysis of selected clones
H Roca et al.
Yeast (Chichester, England), 12(12), 1187-1200 (1996-09-30)
The DEX gene encoding an extracellular dextranase was isolated from the genomic DNA library of Penicillium minioluteum by hybridization using the dextranase cDNA as a probe. Comparison of the gene and cDNA sequences revealed that the DEX gene does not
G S Chaga et al.
Biotechnology and applied biochemistry, 26 ( Pt 1), 7-14 (1997-08-01)
(1) A new concept for producing soluble polymeric affinity ligands is proposed and exemplified. By solid-phase synthesis, an insoluble hydrophilic polymer is converted into an affinity gel. The gel is hydrolytically degraded to water-soluble affinity polymeric ligands which are recovered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service