Merck
  • Alteration of the interconversion of pyruvate and malate in the plastid or cytosol of ripening tomato fruit invokes diverse consequences on sugar but similar effects on cellular organic acid, metabolism, and transitory starch accumulation.
Alteration of the interconversion of pyruvate and malate in the plastid or cytosol of ripening tomato fruit invokes diverse consequences on sugar but similar effects on cellular organic acid, metabolism, and transitory starch accumulation.
Plant physiology (2012-12-20)
Sonia Osorio, José G Vallarino, Marek Szecowka, Shai Ufaz, Vered Tzin, Ruthie Angelovici, Gad Galili, Alisdair R Fernie
ABSTRACT
The aim of this work was to investigate the effect of decreased cytosolic phosphoenolpyruvate carboxykinase (PEPCK) and plastidic NADP-dependent malic enzyme (ME) on tomato (Solanum lycopersicum) ripening. Transgenic tomato plants with strongly reduced levels of PEPCK and plastidic NADP-ME were generated by RNA interference gene silencing under the control of a ripening-specific E8 promoter. While these genetic modifications had relatively little effect on the total fruit yield and size, they had strong effects on fruit metabolism. Both transformants were characterized by lower levels of starch at breaker stage. Analysis of the activation state of ADP-glucose pyrophosphorylase correlated with the decrease of starch in both transformants, which suggests that it is due to an altered cellular redox status. Moreover, metabolic profiling and feeding experiments involving positionally labeled glucoses of fruits lacking in plastidic NADP-ME and cytosolic PEPCK activities revealed differential changes in overall respiration rates and tricarboxylic acid (TCA) cycle flux. Inactivation of cytosolic PEPCK affected the respiration rate, which suggests that an excess of oxaloacetate is converted to aspartate and reintroduced in the TCA cycle via 2-oxoglutarate/glutamate. On the other hand, the plastidic NADP-ME antisense lines were characterized by no changes in respiration rates and TCA cycle flux, which together with increases of pyruvate kinase and phosphoenolpyruvate carboxylase activities indicate that pyruvate is supplied through these enzymes to the TCA cycle. These results are discussed in the context of current models of the importance of malate during tomato fruit ripening.
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, ≥600 units/mg protein (biuret), ammonium sulfate suspension
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)
Sigma-Aldrich
Malic Dehydrogenase (oxalacetate-decarboxylating) from chicken liver, ammonium sulfate suspension, 10-30 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
L-(−)-Malic acid disodium salt, ≥95% (titration)
Sigma-Aldrich
DL-Malic acid, 99%
Sigma-Aldrich
D-(+)-Malic acid, unnatural form, ≥97.0% (T)
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%
Supelco
D-Malic acid, analytical standard
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
Malic Dehydrogenase from bovine heart, ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)