Skip to Content
Merck
All Photos(1)

Key Documents

M5257

Sigma-Aldrich

Malic Dehydrogenase (oxalacetate-decarboxylating) from chicken liver

ammonium sulfate suspension, 10-30 units/mg protein (modified Warburg-Christian)

Synonym(s):

L-Malate:NADP+ oxidoreductase-(oxalacetate-decarboxylating), Malate Dehydrogenase (oxalacetate-decarboxylating), Malic Enzyme

Sign Into View Organizational & Contract Pricing

Select a Size

25 UNITS
$482.00
100 UNITS
$1,590.00

$482.00


Check Cart for Availability


Select a Size

Change View
25 UNITS
$482.00
100 UNITS
$1,590.00

About This Item

CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$482.00


Check Cart for Availability

biological source

chicken liver

form

ammonium sulfate suspension

specific activity

10-30 units/mg protein (modified Warburg-Christian)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Malic dehydrogenase (MDH) exists as two isoforms within eukaryotic cells, one that is expressed in the mitochondria and functions in the TCA cycle and one in the cytoplasm that converts malate from the mitochondria back into oxaloacetate. [1]

Application

Malic dehydrogenase has been used in a study to assess the dietary manganese requirement of juvenile yellow catfish (Pelteobagrus fulvidraco) and effects on whole body mineral composition and hepatic intermediary metabolism. [2] It has also been used in a study to investigate the establishment and biological characterization of a fibroblast cell line from the Langshan chicken. [3]

Biochem/physiol Actions

Expression of malic dehydrogenase in chicken liver is induced upon feeding and decreases in response to starvation. [4]

Unit Definition

One unit will convert 1.0 μmole of L-malate and NADP to pyruvate, CO2, and NADPH per min at pH 7.4 at 25 °C.

Physical form

Suspension in 2.9 M (NH4)2SO4 solution containing 10 mM potassium phosphate, 0.5 mM 2-mercaptoethanol, 10 mM manganese chloride, and 3 mM Na4EDTA, pH 6.0

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Ashton
Archives of biochemistry and biophysics, 345(2), 251-258 (1997-10-06)
NADP-malic enzyme (NADP-ME, EC 1.1.1.40) was purified to near-homogeneity from leaves of the C4 dicot Flaveria bidentis and shown to possess intrinsic NAD-dependent malic enzyme activity. The NAD-dependent activity is optimal at pH 7.5 and in the presence of Mn2+.
Synthesis and Degradation of Malic Enzyme in Chick Liver.
Silpananta, P
The Journal of Biological Chemistry, 264, 5754-5761 (1971)
Dietary manganese requirement of juvenile yellow catfish Pelteobagrus fulvidraco, and effects on whole body mineral composition and hepatic intermediary metabolism
Tan, X., et al.
Aquaculture (Amsterdam, Netherlands), 326, 68-73 (2012)
Establishment and biological characterization of fibroblast cell line from the Langshan chicken
Guan, W., et al.
Cell Proliferation, 42, 157-163 (2010)
K N Rao et al.
Gastroenterology, 113(1), 238-248 (1997-07-01)
We showed previously that the peroxisome proliferators di(2-ethylhexyl)phthalate (DEHP), clofibrate, and 4-chloro-6-(2,3 xylidino)-2-pyrimidinylthio (N-beta-hydroxyl)acetamide (BR931) alter hepatic sex steroid metabolism and receptor expression during induction of hepatic hyperplasia and hepatocellular carcinoma (HCC) in rats. The aim of this study was

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service