Merck
  • Improved Candida methylica formate dehydrogenase fermentation through statistical optimization of low-cost culture media.
Improved Candida methylica formate dehydrogenase fermentation through statistical optimization of low-cost culture media.
Preparative biochemistry & biotechnology (2012-10-04)
Mustafa Kahraman, Emel Bıçakçı Ordu, Koray Yeşiladalı, Nevin Gül Karagüler, Candan Tamerler
ABSTRACT
NAD⁺-dependent formate dehydrogenase (FDH, EC 1.2.1.2) is of use in the regeneration of NAD(P)H coenzymes, and therefore has strong potential for practical application in chemical and medical industries. A low-cost production of recombinant Escherichia coli (E. coli) containing FDH from Candida methylica (cmFDH) was optimized in molasses-based medium by using response surface methodology (RSM) based on central composite design (CCD). The beet molasses as a sole carbon source, (NH₄)₂HPO₄ as a nitrogen and phosphorus source, KH₂PO₄ as a buffer agent, and Mg₂SO₄ · 7H₂O as a magnesium and sulfur source were used as variables in the medium. The optimum medium composition was found to be 34.694 g L⁻¹ of reducing sugar (equivalent to molasses solution), 8.536 g L⁻¹ of (NH₄)₂HPO₄, 3.073 g L⁻¹ of KH₂PO₄, and 1.707 g L⁻¹ of Mg₂SO₄ · 7H₂O. Molasses-based culture medium increased the yield of cmFDH about three times compared to LB medium. The currently developed media has the potential to be used in industrial bioprocesses with low-cost production.
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Formate Dehydrogenase from Candida boidinii, lyophilized powder, 5.0-15.0 units/mg protein
Sigma-Aldrich
Potassium phosphate tribasic, Vetec, reagent grade, 97%
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium phosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Supelco
Ammonium phosphate monobasic, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Ammonium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Sigma-Aldrich
Ammonium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%