Merck
  • METTL3 regulates m6A methylation of PTCH1 and GLI2 in Sonic hedgehog signaling to promote tumor progression in SHH-medulloblastoma.
METTL3 regulates m6A methylation of PTCH1 and GLI2 in Sonic hedgehog signaling to promote tumor progression in SHH-medulloblastoma.
Cell reports (2022-10-27)
Zhi-Wei Zhang, Xufei Teng, Fu Zhao, Chunhui Ma, Jing Zhang, Ling-Feng Xiao, Yaning Wang, Mengqi Chang, Yongji Tian, Chunde Li, Zhang Zhang, Shuhui Song, Wei-Min Tong, Pinan Liu, Yamei Niu
ABSTRACT
SHH subgroup medulloblastoma (SHH-MB) is one of the most common malignant pediatric tumors that arises in the cerebellum. Previously, we showed that RNA m6A methylation participates in regulation of cerebellar development. Here we investigate whether dysregulated m6A methylation contributes to tumorigenesis of SHH-MB. We show that high expression of m6A methyltransferase METTL3 associates with worse survival in the patients with SHH-MB. A large number of hypermethylated transcripts are identified in SHH-MB tumor cells by m6A-seq. We find that METTL3 promotes tumor progression via activating Sonic hedgehog signaling. Mechanistically, METTL3 methylates PTCH1 and GLI2 RNAs and further regulates their RNA stability and translation. Importantly, targeting METTL3 by depleting METTL3 expression or treatment with its catalytic inhibitor STM2457 restrains tumor progression. Collectively, this study shows a critical function for METTL3 and m6A methylation in SHH-MB, indicative of a potential role of METTL3 as therapeutic target in SHH-MB.
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Roche
DNase I recombinant, RNase-free, from bovine pancreas, expressed in Pichia pastoris
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Anti-METTL14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse