Merck
  • Extracellular matrix remodeling through endocytosis and resurfacing of Tenascin-R.
Extracellular matrix remodeling through endocytosis and resurfacing of Tenascin-R.
Nature communications (2021-12-10)
Tal M Dankovich, Rahul Kaushik, Linda H M Olsthoorn, Gabriel Cassinelli Petersen, Philipp Emanuel Giro, Verena Kluever, Paola Agüi-Gonzalez, Katharina Grewe, Guobin Bao, Sabine Beuermann, Hannah Abdul Hadi, Jose Doeren, Simon Klöppner, Benjamin H Cooper, Alexander Dityatev, Silvio O Rizzoli
ABSTRACT
The brain extracellular matrix (ECM) consists of extremely long-lived proteins that assemble around neurons and synapses, to stabilize them. The ECM is thought to change only rarely, in relation to neuronal plasticity, through ECM proteolysis and renewed protein synthesis. We report here an alternative ECM remodeling mechanism, based on the recycling of ECM molecules. Using multiple ECM labeling and imaging assays, from super-resolution optical imaging to nanoscale secondary ion mass spectrometry, both in culture and in brain slices, we find that a key ECM protein, Tenascin-R, is frequently endocytosed, and later resurfaces, preferentially near synapses. The TNR molecules complete this cycle within ~3 days, in an activity-dependent fashion. Interfering with the recycling process perturbs severely neuronal function, strongly reducing synaptic vesicle exo- and endocytosis. We conclude that the neuronal ECM can be remodeled frequently through mechanisms that involve endocytosis and recycling of ECM proteins.
Sigma-Aldrich
Chondroitinase ABC from Proteus vulgaris, BSA free, lyophilized powder, specific activity 50-250 units/mg protein
Sigma-Aldrich
Hyaluronic Acid Binding Protein bovine, lyophilized powder
Sigma-Aldrich
Anti-TGN38 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
Sigma-Aldrich
Lectin from Wisteria floribunda, lyophilized powder
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Anti-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Neurocan antibody, Mouse monoclonal, clone 650.24, purified from hybridoma cell culture
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)