Merck
  • Natural product-based phenols as novel probes for mycobacterial and fungal carbonic anhydrases.
Natural product-based phenols as novel probes for mycobacterial and fungal carbonic anhydrases.
Journal of medicinal chemistry (2011-02-22)
Rohan A Davis, Andreas Hofmann, Asiah Osman, Rebecca A Hall, Fritz A Mühlschlegel, Daniela Vullo, Alessio Innocenti, Claudiu T Supuran, Sally-Ann Poulsen
ABSTRACT
In order to discover novel probes that may help in the investigation and control of infectious diseases through a new mechanism of action, we have evaluated a library of phenol-based natural products (NPs) for enzyme inhibition against four recently characterized pathogen β-family carbonic anhydrases (CAs). These include CAs from Mycobacterium tuberculosis, Candida albicans, and Cryptococcus neoformans as well as α-family human CA I and CA II for comparison. Many of the NPs selectively inhibited the mycobacterial and fungal β-CAs, with the two best performing compounds displaying submicromolar inhibition with a preference for fungal over human CA inhibition of more than 2 orders of magnitude. These compounds provide the first example of non-sulfonamide inhibitors that display β over α CA enzyme selectivity. Structural characterization of the library compounds in complex with human CA II revealed a novel binding mode whereby a methyl ester interacts via a water molecule with the active site zinc.
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
2-(4-Hydroxyphenyl)ethanol, 98%
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
Sulfanilamide, ≥98%
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Supelco
2-(4-Hydroxyphenyl)ethanol, analytical standard
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard