Merck
  • Extracellular tau induces microglial phagocytosis of living neurons in cell cultures.
Extracellular tau induces microglial phagocytosis of living neurons in cell cultures.
Journal of neurochemistry (2019-12-14)
Katryna Pampuscenko, Ramune Morkuniene, Tomas Sneideris, Vytautas Smirnovas, Rima Budvytyte, Gintaras Valincius, Guy C Brown, Vilmante Borutaite
ABSTRACT
Tau is a microtubule-associated protein, found at high levels in neurons, and its aggregation is associated with neurodegeneration. Recently, it was found that tau can be actively secreted from neurons, but the effects of extracellular tau on neuronal viability are unclear. In this study, we investigated whether extracellular tau2N4R can cause neurotoxicity in primary cultures of rat brain neurons and glial cells. Cell cultures were examined for neuronal loss, death, and phosphatidylserine exposure, as well as for microglial phagocytosis by fluorescence microscopy. Aggregation of tau2N4R was assessed by atomic force microscopy. We found that extracellular addition of tau induced a gradual loss of neurons over 1-2 days, without neuronal necrosis or apoptosis, but accompanied by proliferation of microglia in the neuronal-glial co-cultures. Tau addition caused exposure of the 'eat-me' signal phosphatidylserine on the surface of living neurons, and this was prevented by elimination of the microglia or by inhibition of neutral sphingomyelinase. Tau also increased the phagocytic activity of pure microglia, and this was blocked by inhibitors of neutral sphingomyelinase or protein kinase C. The neuronal loss induced by tau was prevented by inhibitors of neutral sphingomyelinase, protein kinase C or the phagocytic receptor MerTK, or by eliminating microglia from the cultures. The data suggest that extracellular tau induces primary phagocytosis of stressed neurons by activated microglia, and identifies multiple ways in which the neuronal loss induced by tau can be prevented.
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
GSK-3β Inhibitor I, The GSK-3β Inhibitor I, also referenced under CAS 327036-89-5, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
L-Leucine methyl ester hydrochloride, 98%
Sigma-Aldrich
GW4869, ≥90% (NMR)
Sigma-Aldrich
Gö 6983, ≥97%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ro-32-0432, A selective cell-permeable protein kinase C inhibitor.