Merck
  • Drug design, in vitro pharmacology, and structure-activity relationships of 3-acylamino-2-aminopropionic acid derivatives, a novel class of partial agonists at the glycine site on the N-methyl-D-aspartate (NMDA) receptor complex.
Drug design, in vitro pharmacology, and structure-activity relationships of 3-acylamino-2-aminopropionic acid derivatives, a novel class of partial agonists at the glycine site on the N-methyl-D-aspartate (NMDA) receptor complex.
Journal of medicinal chemistry (2009-08-01)
Stephan Urwyler, Philipp Floersheim, Bernard L Roy, Manuel Koller
ABSTRACT
Retaining agonistic activity at the glycine coagonist site of the NMDA receptor in molecules derived from glycine or d-serine has proven to be difficult because in the vicinity of the alpha-amino acid group little substitution is tolerated. We have solved this problem by replacing the hydroxy group of d-serine with an amido group, thus keeping the hydrogen donor function and allowing for further substitution and exploration of the adjacent space. Heterocyclic substitutions resulted in a series of 3-acylamino-2-aminopropionic acid derivatives, with high affinities in a binding assay for the glycine site. In a functional assay assessing the activation of the glycine site, these compounds displayed a wide range of intrinsic efficacies, from antagonism to a high degree of partial agonism. Structure-activity relationships reveal that lipophilic substituents, presumably filling an additional hydrophobic pocket, are accepted by the glycine site, provided that they are separated from the alpha-amino acid group by a short linker.
Sigma-Aldrich
D-Alanine, ≥98% (HPLC)
Sigma-Aldrich
Cycloleucine, 97%
Sigma-Aldrich
Glycine 1 M solution
Sigma-Aldrich
D-Cycloserine, synthetic
Sigma-Aldrich
D-Cycloserine
Sigma-Aldrich
D-Serine, ≥98% (TLC)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
D-2-Aminobutyric acid, 98%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
SAFC
Glycine
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
1-Aminocyclopropanecarboxylic acid, ≥98% (TLC), powder
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%