Merck
Structure and conformational dynamics of trichothecene mycotoxins.
Journal of natural products (2008-03-11)
Wayne E Steinmetz, Paul Robustelli, Eric Edens, David Heineman
ABSTRACT
A combination of NMR spectroscopy and molecular modeling has been employed to characterize the conformation and dynamics of the macrolide ring in verrucarin A and roridin A, two closely related toxins in the trichothecene mycotoxin family. Longitudinal carbon-13 relaxation times demonstrate the relative flexibility of the macrolide ring. The calculations, NOEs, and scalar vicinal coupling constants show that verrucarin A in CDCl 3 and CD 2Cl 2 predominantly adopts a single, well-defined conformation that matches the crystal structure. In contrast, roridin A is present as a mixture of two conformers.
Product Description
Nivalenol in acetonitrile, IRMM®, certified reference material
4-Deoxynivalenol in acetonitrile, IRMM®, certified reference material
Supelco
T2-Toxin solution, 100 μg/mL in acetonitrile, analytical standard
Supelco
Deoxynivalenol solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Deoxynivalenol
Sigma-Aldrich
T-2 Toxin, from Fusarium sp., powder, ≥98% (HPLC)
Supelco
Nivalenol solution, 100 μg/mL in acetonitrile, analytical standard
Supelco
3-Acetyldeoxynivalenol solution, ~100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
15-O-Acetyl-4-deoxynivalenol from Fusarium graminearum
Sigma-Aldrich
3-Acetyldeoxynivalenol, from Fusarium roseum