Merck
Hologram QSAR model for the prediction of human oral bioavailability.
Bioorganic & medicinal chemistry (2007-09-18)
Tiago L Moda, Carlos A Montanari, Adriano D Andricopulo
ABSTRACT
A drug intended for use in humans should have an ideal balance of pharmacokinetics and safety, as well as potency and selectivity. Unfavorable pharmacokinetics can negatively affect the clinical development of many otherwise promising drug candidates. A variety of in silico ADME (absorption, distribution, metabolism, and excretion) models are receiving increased attention due to a better appreciation that pharmacokinetic properties should be considered in early phases of the drug discovery process. Human oral bioavailability is an important pharmacokinetic property, which is directly related to the amount of drug available in the systemic circulation to exert pharmacological and therapeutic effects. In the present work, hologram quantitative structure-activity relationships (HQSAR) were performed on a training set of 250 structurally diverse molecules with known human oral bioavailability. The most significant HQSAR model (q(2)=0.70, r(2)=0.93) was obtained using atoms, bond, connection, and chirality as fragment distinction. The predictive ability of the model was evaluated by an external test set containing 52 molecules not included in the training set, and the predicted values were in good agreement with the experimental values. The HQSAR model should be useful for the design of new drug candidates having increased bioavailability as well as in the process of chemical library design, virtual screening, and high-throughput screening.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Saccharin, ≥99%
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
Sigma-Aldrich
(S)-(+)-Ketoprofen, 99%
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Chlorambucil
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV)
Sigma-Aldrich
19-Norethindrone, ≥98%, powder
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Supelco
Digoxin, analytical standard
Sigma-Aldrich
Glipizide, solid
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Supelco
D(−)-Norgestrel, analytical standard
Sigma-Aldrich
2-Propylpentanoic acid