Skip to Content
Merck
All Photos(1)

Key Documents

P6273

Sigma-Aldrich

2-Propylpentanoic acid

Synonym(s):

Valproic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$281.00

$281.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$281.00

About This Item

Linear Formula:
(CH3CH2CH2)2CHCO2H
CAS Number:
Molecular Weight:
144.21
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

$281.00


Check Cart for Availability

Request a Bulk Order

refractive index

n20/D 1.425 (lit.)

bp

220 °C (lit.)

solubility

H2O: slightly soluble
alcohol: soluble

density

0.9 g/mL at 25 °C (lit.)

lipid type

saturated FAs

SMILES string

CCCC(C(O)=O)CCC

InChI

1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

NIJJYAXOARWZEE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

2-Propylpentanoic acid has been used as a supplement in mouse embryonic fibroblast - conditioned medium (MEF-CM) to feed the cells.[1]

Biochem/physiol Actions

2-Propylpentanoic acid is a short-chain fatty acid and potentially inhibits histone deacetylase action. It is known to promote tumor cell toxicity and increase susceptibility to radiation.[2] 2-Propylpentanoic acid elevates the levels of γ-aminobutyric acid in the brain. Reduced central nervous system action is observed in overdosage of 2-propylpentanoic acid.[3] It enhance the inhibitory action of γ-aminobutyric acid (GABA) by blocking voltage-dependent sodium channel.[4]
Anticonvulsant that also has efficacy as a mood stabilizer in bipolar disorder

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1A - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient generation of nonhuman primate induced pluripotent stem cells
Zhong B, et al.
Stem Cells and Development, 20(5), 795-807 (2010)
3-Heptanone as a potential new marker for valproic acid therapy
Erhart S, et al.
Journal of Breath Research, 3(1), 016004-016004 (2009)
Histone deacetylase inhibitor, 2-propylpentanoic acid, increases the chemosensitivity and radiosensitivity of human glioma cell linesin vitro
Cuijie S, et al.
Chinese Medical Journal (English Edition), 125(24), 4338-4343 (2012)
Xi Chen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(21), 9202-9213 (2013-05-24)
Progranulin (PGRN) is a secreted glycoprotein expressed in neurons and glia that is implicated in neuronal survival on the basis that mutations in the GRN gene causing haploinsufficiency result in a familial form of frontotemporal dementia (FTD). Recently, a direct
P C Ho et al.
The pharmacogenomics journal, 3(6), 335-342 (2003-11-05)
The present study investigated the effect of cytochrome P450 2C9 (CYP2C9) genetic polymorphism on the biotransformation of valproic acid (VPA) to its hepatotoxic metabolite, 4-ene-VPA, and compared that to the formation of the inactive 4-OH-VPA and 5-OH-VPA. cDNA-expressed CYP2C9(*)2 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service