Merck
  • Increased plasma bradykinin level is associated with cognitive impairment in Alzheimer's patients.
Increased plasma bradykinin level is associated with cognitive impairment in Alzheimer's patients.
Neurobiology of disease (2020-03-17)
Pradeep K Singh, Zu-Lin Chen, Dhiman Ghosh, Sidney Strickland, Erin H Norris
ABSTRACT
Alzheimer's disease (AD) is characterized by the presence of proteinaceous brain deposits, brain atrophy, vascular dysfunction, and chronic inflammation. Along with cerebral inflammation, peripheral inflammation is also evident in many AD patients. Bradykinin, a proinflammatory plasma peptide, is also linked to AD pathology. For example, bradykinin infusion into the hippocampus causes learning and memory deficits in rats, and blockade of the bradykinin receptor lessens cognitive impairment in AD mouse models. Even though it has been hypothesized that plasma bradykinin could contribute to inflammation in AD, the level of plasma bradykinin and its association with beta-amyloid (Aβ) pathology in AD patients had not been explored. Here, we assessed plasma bradykinin levels in AD patients and age-matched non-demented (ND) control individuals. We found significantly elevated plasma bradykinin levels in AD patients compared to ND subjects. Additionally, changes in plasma bradykinin levels were more profound in many AD patients with severe cognitive impairment, suggesting that peripheral bradykinin could play a role in dementia most likely via inflammation. Bradykinin levels in the cerebrospinal fluid (CSF) were reduced in AD patients and exhibited an inverse correlation with the CSF Aβ40/Aβ42 ratio. We also report that bradykinin interacts with the fibrillar form of Aβ and co-localizes with Aβ plaques in the post-mortem human AD brain. These findings connect the peripheral inflammatory pathway to cerebral abnormalities and identify a novel mechanism of inflammatory pathology in AD.
Sigma-Aldrich
Angiotensin I Converting Enzyme (ACE) Activity Assay Kit (Fluorometric), sufficient for 200 fluorometric tests
Pricing and availability is not currently available.
Sigma-Aldrich
Human Kininogen ELISA Kit, for cell culture supernatants, plasma, and serum samples
Pricing and availability is not currently available.