Skip to Content
Merck
All Photos(1)

Key Documents

RAB1068

Sigma-Aldrich

Human Kininogen ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,400.00

$1,400.00


Check Cart for Availability


Select a Size

Change View
96 WELLS
$1,400.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,400.00


Check Cart for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum sample(s)
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 24 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KNG1(3827)

General description

This ELISA antibody pair detects human Kininogen. Other species are not determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tae Yeul Kim et al.
Journal of thrombosis and thrombolysis, 46(3), 379-385 (2018-08-14)
Leukemic cells release their nuclear contents into the extracellular space upon activation. The released nuclear contents, called extracellular traps, can activate the contact system of coagulation. This study accessed the extent of contact system activation, the levels of extracellular traps
Ying Zhou et al.
International journal of oncology, 55(1), 131-141 (2019-05-07)
Bradykinin (BK) is one of the kinin peptides and preferentially binds to bradykinin B2 receptor (BDKRB2). A recent study indicated that BK played an important role in the occurrence and progression of cancer. In this study, we evaluated the serum BK levels in
Weiqing Fu et al.
Food & function, 11(4), 3645-3656 (2020-04-17)
In this study, the blood pressure-lowering effects of Trichiurus lepturus myosin hydrolysate (TMH) and its possible mechanism were investigated in spontaneously hypertensive rats (SHRs). After gavage administration of TMH for 4 h, systolic blood pressure (SBP) was significantly decreased in
Ruwan Gunaratne et al.
Nature biotechnology, 36(7), 606-613 (2018-06-05)
Unfractionated heparin (UFH), the standard anticoagulant for cardiopulmonary bypass (CPB) surgery, carries a risk of post-operative bleeding and is potentially harmful in patients with heparin-induced thrombocytopenia-associated antibodies. To improve the activity of an alternative anticoagulant, the RNA aptamer 11F7t, we
Jung Eun Park et al.
Biomarker research, 9(1), 5-5 (2021-01-11)
Alzheimer's disease (AD) is the most common cause of dementia and most of AD patients suffer from vascular abnormalities and neuroinflammation. There is an urgent need to develop novel blood biomarkers capable of diagnosing Alzheimer's disease (AD) at very early

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service