Merck
  • The effects of human immunoglobulin G on enhancing tissue protection and neurobehavioral recovery after traumatic cervical spinal cord injury are mediated through the neurovascular unit.
The effects of human immunoglobulin G on enhancing tissue protection and neurobehavioral recovery after traumatic cervical spinal cord injury are mediated through the neurovascular unit.
Journal of neuroinflammation (2019-07-11)
Jonathon Chon Teng Chio, Jian Wang, Anna Badner, James Hong, Vithushan Surendran, Michael G Fehlings
ABSTRACT
Spinal cord injury (SCI) is a condition with few effective treatment options. The blood-spinal cord barrier consists of pericytes, astrocytes, and endothelial cells, which are collectively termed the neurovascular unit. These cells support spinal cord homeostasis by expressing tight junction proteins. Physical trauma to the spinal cord disrupts the barrier, which leads to neuroinflammation by facilitating immune cell migration to the damaged site in a process involving immune cell adhesion. Immunosuppressive strategies, including methylprednisolone (MPSS), have been investigated to treat SCI. However, despite some success, MPSS has the potential to increase a patient's susceptibility to wound infection and impaired wound healing. Hence, immunomodulation may be a more attractive approach than immunosuppression. Approved for modulating neuroinflammation in certain disorders, including Guillain-Barre syndrome, intravenous administration of human immunoglobulin G (hIgG) has shown promise in the setting of experimental SCI, though the optimal dose and mechanism of action remain undetermined. Female adult Wistar rats were subjected to moderate-severe clip compression injury (35 g) at the C7-T1 level and randomized to receive a single intravenous (IV) bolus of hIgG (0.02, 0.2, 0.4, 1, 2 g/kg), MPSS (0.03 g/kg), or control buffer at 15 min post-SCI. At 24 h and 6 weeks post-SCI, molecular, histological, and neurobehavioral effects of hIgG were analyzed. At 24 h post-injury, human immunoglobulin G co-localized with spinal cord pericytes, astrocytes, and vessels. hIgG (2 g/kg) protected the spinal cord neurovasculature after SCI by increasing tight junction protein expression and reducing inflammatory enzyme expression. Improvements in vascular integrity were associated with changes in spinal cord inflammation. Interestingly, hIgG (2 g/kg) increased serum expression of inflammatory cytokines and co-localized (without decreasing protein expression) with spinal cord vascular cell adhesion molecule-1, a protein used by immune cells to enter into inflamed tissue. Acute molecular benefits of hIgG (2 g/kg) led to greater tissue preservation, functional blood flow, and neurobehavioral recovery at 6 weeks post-SCI. Importantly, the effects of hIgG (2 g/kg) were superior to control buffer and hIgG (0.4 g/kg), and comparable with MPSS (0.03 g/kg). hIgG (2 g/kg) is a promising therapeutic approach to mitigate secondary pathology in SCI through antagonizing immune cell infiltration at the level of the neurovascular unit.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-MMP-9 Antibody, Catalytic domain, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Nitric Oxide Synthase II Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-ZO-1 Antibody, clone R40.76, clone R40.76, from rat