Merck
  • Modelling retention and peak shape of small polar solutes analysed by nano-HPLC using methacrylate-based monolithic columns.
Modelling retention and peak shape of small polar solutes analysed by nano-HPLC using methacrylate-based monolithic columns.
Analytica chimica acta (2019-09-29)
J A Navarro-Huerta, E J Carrasco-Correa, J R Torres-Lapasió, J M Herrero-Martínez, M C García-Alvarez-Coque
ABSTRACT
The development of methacrylate-based monolithic columns was studied for the separation of pharmaceutical hydrophilic compounds in nano-liquid chromatography. The selected polymerisation mixture consisted of 7.5% hexyl methacrylate, 4.5% methacrylic acid and 18.0% ethylene dimethacrylate (w/w), in a binary porogenic solvent (35:35 w/w 1-propanol/1,4-butanediol). The polymer synthesised with this mixture has a good permeability, not excessive back-pressure, and reasonable retention times for polar and non-polar solutes. Monolithic columns (12 cm total capillary length, 100 μm i.d.), prepared with this mixture, were able to produce hydrogen bonding and electrostatic interactions, giving rise to promising separations. To evaluate the chromatographic system, alkylbenzenes (neutral and hydrophobic compounds) and sulphonamides (hydrophilic drugs) were assayed. To optimise the chromatographic mobile phase in isocratic elution and characterise the retention mechanism for a mixture of eight sulphonamides, the performance of several mathematic models was checked in the description of retention. The behaviour of the monolithic capillary column was compared, in terms of selectivity and peak shape, to that obtained with a C18 column (9 cm × 4.6 mm i.d., 5 μm particle size) using a conventional HPLC equipment. The results revealed substantial differences in the interactions established for sulphonamides between the monolithic and C18 columns.
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor