Merck
  • Repression of Transcription at DNA Breaks Requires Cohesin throughout Interphase and Prevents Genome Instability.
Repression of Transcription at DNA Breaks Requires Cohesin throughout Interphase and Prevents Genome Instability.
Molecular cell (2018-12-18)
Cornelia Meisenberg, Sarah I Pinder, Suzanna R Hopkins, Sarah K Wooller, Graeme Benstead-Hume, Frances M G Pearl, Penny A Jeggo, Jessica A Downs
ABSTRACT
Cohesin subunits are frequently mutated in cancer, but how they function as tumor suppressors is unknown. Cohesin mediates sister chromatid cohesion, but this is not always perturbed in cancer cells. Here, we identify a previously unknown role for cohesin. We find that cohesin is required to repress transcription at DNA double-strand breaks (DSBs). Notably, cohesin represses transcription at DSBs throughout interphase, indicating that this is distinct from its known role in mediating DNA repair through sister chromatid cohesion. We identified a cancer-associated SA2 mutation that supports sister chromatid cohesion but is unable to repress transcription at DSBs. We further show that failure to repress transcription at DSBs leads to large-scale genome rearrangements. Cancer samples lacking SA2 display mutational patterns consistent with loss of this pathway. These findings uncover a new function for cohesin that provides insights into its frequent loss in cancer.
Sigma-Aldrich
Hoechst 33258 solution, 1 mg/mL in H2O, ≥98.0% (HPLC)
Sigma-Aldrich
bisBenzimide H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Goat Anti-Rat IgG Antibody, HRP conjugate, Chemicon®, from goat
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-CTCF Antibody, serum, Upstate®
Sigma-Aldrich
Anti-ubiquityl-Histone H2A Antibody, clone E6C5, clone E6C5, Upstate®, from mouse
Sigma-Aldrich
Anti-BAF180 Antibody, from rabbit