Merck
  • A Protective Role for the Lectin CD169/Siglec-1 against a Pathogenic Murine Retrovirus.
A Protective Role for the Lectin CD169/Siglec-1 against a Pathogenic Murine Retrovirus.
Cell host & microbe (2019-01-01)
Pradeep D Uchil, Ruoxi Pi, Kelsey A Haugh, Mark S Ladinsky, John D Ventura, Brad S Barrett, Mario L Santiago, Pamela J Bjorkman, George Kassiotis, Xaver Sewald, Walther Mothes
ABSTRACT
Lymph- and blood-borne retroviruses exploit CD169/Siglec-1-mediated capture by subcapsular sinus and marginal zone metallophilic macrophages for trans-infection of permissive lymphocytes. However, the impact of CD169-mediated virus capture on retrovirus dissemination and pathogenesis in vivo is unknown. In a murine model of the splenomegaly-inducing retrovirus Friend virus complex (FVC) infection, we find that while CD169 promoted draining lymph node infection, it limited systemic spread to the spleen. At the spleen, CD169-expressing macrophages captured incoming blood-borne retroviruses and limited their spread to the erythroblasts in the red pulp where FVC manifests its pathogenesis. CD169-mediated retroviral capture activated conventional dendritic cells 1 (cDC1s) and promoted cytotoxic CD8+ T cell responses, resulting in efficient clearing of FVC-infected cells. Accordingly, CD169 blockade led to higher viral loads and accelerated death in susceptible mouse strains. Thus, CD169 plays a protective role during FVC pathogenesis by reducing viral dissemination to erythroblasts and eliciting an effective cytotoxic T lymphocyte response via cDC1s.
Sigma-Aldrich
3-Amino-4-methoxybenzenesulfonic acid, 98%
Sigma-Aldrich
FTY720, ≥98% (HPLC)
Sigma-Aldrich
Phorbol 12-Myristate 13-Acetate
Sigma-Aldrich
Red Blood Cell Lysing Buffer Hybri-Max, liquid, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
Mix-n-Stain CF 647 Antibody Labeling Kit (50-100μg)
Sigma-Aldrich
Mix-n-Stain CF 488A Antibody Labeling Kit (50-100μg)