Merck
c-Jun-mediated anticancer mechanisms of tylophorine.
Carcinogenesis (2013-02-07)
Cheng-Wei Yang, Yue-Zhi Lee, Hsing-Yu Hsu, Chia-Mao Wu, Hwan-You Chang, Yu-Sheng Chao, Shiow-Ju Lee
ABSTRACT
Tylophorine, a phenanthroindolizidine alkaloid, is the major medicinal constituent of herb Tylophora indica. Tylophorine treatment increased the accumulation of c-Jun protein, a component of activator protein 1 (AP1), in carcinoma cells. An in vitro kinase assay revealed that the resultant c-Jun phosphorylation was primarily mediated via activated c-Jun N-terminal protein kinase (JNK). Moreover, flow cytometry indicated that ectopically overexpressed c-Jun in conjunction with tylophorine significantly increased the number of carcinoma cells that were arrested at the G1 phase. The tylophorine-mediated downregulation of cyclin A2 protein levels is known to be involved in the primary G1 arrest. Chromatin immunoprecipitation and reporter assays revealed that tylophorine enhanced the c-Jun downregulation of the cyclin A2 promoter activity upon increased binding of c-Jun to the deregulation AP1 site and decreased binding to the upregulation activating transcription factor (ATF) site in the cyclin A2 promoter, thereby reducing cyclin A2 expression. Further, biochemical studies using pharmacological inhibitors and RNA silencing approaches demonstrated that tylophorine-mediated elevation of the c-Jun protein level occurs primarily via two discrete prolonged signaling pathways: (i) the NF-κB/PKCδ_(MKK4)_JNK cascade, which phosphorylates c-Jun and increases its stability by slowing its ubiquitination, and (ii) the PI3K_PDK1_PP2A_eEF2 cascade, which sustains eukaryotic elongation factor 2 (eEF2) activity and thus c-Jun protein translation. To the best of our knowledge, this report is the first to demonstrate the involvement of c-Jun in the anticancer activity of tylophorine and the release of c-Jun translation from a global translational blockade via the PI3K_PDK1_eEF2 signaling cascade.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Okadaic Acid, Prorocentrum sp., Okadaic Acid, CAS 78111-17-8, is a highly potent inhibitor of protein phosphatase 1 (IC₅₀ = 10-15 nM) and 2A (IC₅₀ = 0.1 nM).
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Sigma-Aldrich
EZ-Magna ChIP® G - Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic G beads. Control primers included.
Sigma-Aldrich
IKK-2 Inhibitor V, The IKK-2 Inhibitor V, also referenced under CAS 978-62-1, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.