
Monoclonal Anti-HIF-1α antibody produced in mouse

Synonym(s):
Anti-Hypoxia Inducible Factor 1, α subunit, Anti-ARNT interacting protein, Anti-HIF1 alpha, ESEE122, Anti-HIF1A, Anti-Hypoxia inducible factor 1 alpha, Anti-MOP1, Anti-PASD8
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing