Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
KDS Legato
™
270P series syringe pump, push/pull, programmable
Synonym(s):
KDS Legato
™
270P series syringe pump, push/pull, programmable
Products
Product Category
pumps (2)
Manufacturer
Volume (ml)
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
Z569623
KDS Legato
™
270P series syringe pump, push/pull, programmable,
AC/DC input 100 - 240 V AC, US 3-pin plug
Expand
View Pricing
Z740541
KDS Legato
™
270P series syringe pump, push/pull, programmable,
AC/DC input 100 - 240 V AC, EuroPlug (flat 2-pin)
Expand
View Pricing