Skip to Content
Merck
Ammonium hydrogen carbonate for HPLC LiChropur™

Ammonium hydrogen carbonate

Ammonium hydrogen carbonate for HPLC LiChropur™
Synonym(s):
Ammonium hydrogen carbonate
Linear Formula:
(NH4)HCO3
CAS Number:
Molecular Weight:
79.06

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Technique
Showing 1-2 of 2