Skip to Content
Merck
All Photos(1)

Key Documents

V900882

Sigma-Aldrich

Diethyl pyrocarbonate

Vetec, reagent grade, ≥97%

Synonym(s):

DEP, DEPC, Diethyl dicarbonate, Diethyl oxydiformate, Ethoxyformic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(COOC2H5)2
CAS Number:
Molecular Weight:
162.14
Beilstein:
637031
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥97%

form

liquid

mol wt

162,14 g/mol

refractive index

n20/D 1.398 (lit.)

bp

93-94 °C/18 mmHg (lit.)

density

1.101 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)OC(=O)OCC

InChI

1S/C6H10O5/c1-3-9-5(7)11-6(8)10-4-2/h3-4H2,1-2H3

InChI key

FFYPMLJYZAEMQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Modification reagent for His and Tyr residues in proteins. Robust probe for structural disruptions in dsDNA, reacting with fully or partially unstacked bases.

Biochem/physiol Actions

Inactivates RNase in solution at about 0.1% (v/v), thus protecting RNA against degradation.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Martineau et al.
Applied and environmental microbiology, 81(15), 5003-5014 (2015-05-17)
Hyphomicrobium spp. are commonly identified as major players in denitrification systems supplied with methanol as a carbon source. However, denitrifying Hyphomicrobium species are poorly characterized, and very few studies have provided information on the genetic and physiological aspects of denitrification
Jonathan L Hess et al.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics, 168B(1), 14-35 (2014-12-20)
Advances in molecular genetics, fueled by the results of large-scale genome-wide association studies, meta-analyses, and mega-analyses, have provided the means of identifying genetic risk factors for human disease, thereby enriching our understanding of the functionality of the genome in the
Zdenka O Kyjovska et al.
Mutagenesis, 30(4), 499-507 (2015-03-17)
We investigated the inflammatory response, acute phase response and genotoxic effect of diesel exhaust particles (DEPs, NIST1650b) following a single intratracheal instillation. C57BL/6J BomTac mice received 18, 54 or 162 µg/mouse and were killed 1, 3 and 28 days post-exposure.
Simona Portaro et al.
Neuromolecular medicine, 17(3), 285-296 (2015-05-27)
Myotonia congenita (MC) is an inherited muscle disease characterized by impaired muscle relaxation after contraction, resulting in muscle stiffness. Both recessive (Becker's disease) or dominant (Thomsen's disease) MC are caused by mutations in the CLCN1 gene encoding the voltage-dependent chloride
Aidong Chen et al.
The Journal of physiology, 592(16), 3523-3536 (2014-06-29)
Circulating Ang II activates an aldosterone-mineralocorticoid receptor (MR) - angiotensin II (Ang II) - angiotensin type 1 receptor (AT1R) pathway in the hypothalamus. To obtain insights into the actual neuronal projections involved, adeno-associated virus carrying small interfering RNA against either

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service