Skip to Content
Merck
All Photos(1)

Key Documents

V900751

Sigma-Aldrich

Pentafluorophenol

Vetec, reagent grade, 98%

Synonym(s):

2,3,4,5,6-Pentafluorophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5OH
CAS Number:
Molecular Weight:
184.06
Beilstein:
1912584
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

bp

143 °C (lit.)

mp

34-36 °C (lit.)

SMILES string

Oc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C6HF5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H

InChI key

XBNGYFFABRKICK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin A Ashu-Arrah et al.
Journal of chromatography. A, 1273, 34-43 (2013-01-02)
Pentafluorophenyl and phenyl silica stationary phases offer alternative selectivity compared to alkyl bonded C₁₈ and C₈ stationary phases, through other interactions such as π-π interactions, dipole-dipole and hydrogen bond interactions. Pentafluorophenyl and phenyl silica bonded stationary phases were efficiently prepared
Lisbeth Grøndahl et al.
Biomacromolecules, 6(4), 2197-2203 (2005-07-12)
Poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBV) melt processed disks and solvent cast films were modified by graft co-polymerization with acrylic acid (AAc) in methanol solution at ambient temperature using gamma irradiation (dose rate of 4.5 kGy/h). To assess the presence of carboxylic acid groups
Daniel P Sanders et al.
Journal of the American Chemical Society, 132(42), 14724-14726 (2010-10-05)
An improved two-step synthetic route to functionalized cyclic carbonate monomers that features a novel cyclic carbonate intermediate with an active pentafluorophenyl ester group (MTC-OPhF(5)) has been developed. The versatile pentafluorophenyl ester intermediate can be synthesized on the gram to kilogram
L K Shao et al.
Analytical chemistry, 69(11), 2008-2016 (1997-06-01)
Baseline separation of 15 taxanes including paclitaxel (Taxol) was achieved on pentafluorophenyl (PFP) HPLC columns. Methods using aqueous acetonitrile gradients on each of two commercial PFP columns were developed that are suitable for the determination of potency, content uniformity, and
Rens J Götz et al.
Chemical communications (Cambridge, England), (29)(29), 3384-3386 (2008-07-18)
The rational design of a ligand containing two electron-poor pi-rings, i.e. a triazine and a pentafluorophenoxy groups, has allowed the preparation of a copper complex where both the anticipated anion...pi interactions are present.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service