Skip to Content
Merck
All Photos(1)

Key Documents

V900670

Sigma-Aldrich

Tyramine

Vetec, reagent grade, 99%

Synonym(s):

2-(4-Hydroxyphenyl)ethylamine, 4-(2-Aminoethyl)phenol, 4-Hydroxyphenethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
137.18
Beilstein:
1099914
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

bp

175-181 °C/8 mmHg (lit.)

mp

160-162 °C (lit.)

SMILES string

NCCc1ccc(O)cc1

InChI

1S/C8H11NO/c9-6-5-7-1-3-8(10)4-2-7/h1-4,10H,5-6,9H2

InChI key

DZGWFCGJZKJUFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Can enter catecholaminergic terminals and be released as a false transmitter.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzan El-Akaad et al.
Scientific reports, 10(1), 14479-14479 (2020-09-04)
This manuscript reports on the development of a capacitive sensor for the detection of imidacloprid (IMD) in water samples based on molecularly imprinted polymers (MIPs). MIPs used as recognition elements were synthesized via a photo-initiated emulsion polymerization. The particles were
Giris Jacob et al.
Circulation, 107(19), 2475-2479 (2003-04-23)
Despite the widespread use of tyramine as a pharmacological tool to assess the effects of norepinephrine release from sympathetic nerve terminals, its vascular effects are not adequately characterized. In particular, previous results indicate that intravenous tyramine produces little if any
Stephan A Chalon et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 28(9), 1685-1693 (2003-06-05)
Evidence suggests that compounds that increase the synaptic availability of more than one neurotransmitter have greater efficacy in the treatment of depression than single-acting drugs. Preclinical studies indicate that duloxetine acts to inhibit serotonin (5-HT) and norepinephrine (NE) transporters. The
Thomas Roeder
Annual review of entomology, 50, 447-477 (2004-09-10)
Octopamine (OA) and tyramine (TA) are the invertebrate counterparts of the vertebrate adrenergic transmitters. They are decarboxylation products of the amino acid tyrosine, with TA as the biological precursor of OA. Nevertheless, both compounds are independent neurotransmitters that act through
Moussa B H Youdim et al.
Neurotoxicology, 25(1-2), 243-250 (2003-12-31)
The major side effect with the use of first generation of non selective monoamine oxidase (MAO) inhibitors as neuropsychiatric drugs was what became known as the "cheese reaction". Namely, potentiation of sympathomimetic activity of ingested tyramine present in cheese and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service