Skip to Content
Merck
All Photos(1)

Documents

V900660

Sigma-Aldrich

Indoline

Vetec, reagent grade, 98%

Synonym(s):

1-Azaindan, 2,3-Dihydroindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N
CAS Number:
Molecular Weight:
119.16
Beilstein:
111915
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

refractive index

n20/D 1.592 (lit.)

bp

220-221 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

C1Cc2ccccc2N1

InChI

1S/C8H9N/c1-2-4-8-7(3-1)5-6-9-8/h1-4,9H,5-6H2

InChI key

LPAGFVYQRIESJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Inhibitors of NOD1-Induced Nuclear Factor-κB Activation
  • Sphingosine-1-phosphate 4(S1P4) receptor antagonists
  • Cytotoxic cell cycle inhibitors
  • 2-Aminopyridines
  • PET agent for imaging of protein kinase C (PKC)
  • Sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes
  • α4β2-Nicotinic acetylcholine receptor-selective partial agonists
  • mGlu4 positive allosteric modulators
  • Bacterial biofilm inhibitors
  • Serotonin 5-HT6 receptor antagonists

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiharu Noji et al.
Organic letters, 15(8), 1946-1949 (2013-04-04)
A benzyne-mediated synthesis of substituted indolines and carbazoles was developed. The reaction includes generation of benzyne using Mg(TMP)2·2LiCl as a base, cyclization, and trapping the resulting organomagnesium intermediate with an electrophile to provide a series of substituted indolines and carbazoles
Manas K Ghorai et al.
The Journal of organic chemistry, 78(8), 3867-3878 (2013-04-04)
A practical approach for the synthesis of 3-substituted indolines via regio- and stereoselective SN2-type ring-opening of 2-(2-halophenyl)-N-tosylaziridines with heteroatomic nucleophiles (O, N, and S) followed by palladium-catalyzed intramolecular C-N cyclization is reported in excellent yields (up to >99%) and enantiomeric
Tanguy Saget et al.
Organic letters, 15(6), 1354-1357 (2013-03-05)
The synthesis of cyclopropyl spiroindolines is described using an intramolecular palladium(0)-catalyzed C-H functionalization of a methine C(sp(3))-H bond. This transformation can be coupled with intermolecular Suzuki couplings or direct arylations of heteroaromatics to access functionalized indoline scaffolds in a single
A diversity-oriented approach to spiroindolines: post-Ugi gold-catalyzed diastereoselective domino cyclization.
Sachin G Modha et al.
Angewandte Chemie (International ed. in English), 51(38), 9572-9575 (2012-08-22)
Ke Chen et al.
Organic & biomolecular chemistry, 10(32), 6600-6606 (2012-07-17)
A palladacycle-catalyzed tandem Heck-intramolecular aza-Michael reaction protocol has been developed for the one-pot synthesis of 1-substituted isoindolines from N-unprotected 2-bromobenzylamines and acrylates with high yields.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service