Skip to Content
Merck
All Photos(3)

Key Documents

V900648

Sigma-Aldrich

Ethylenediamine dihydrochloride

Vetec, reagent grade, 98%

Synonym(s):

1,2-Diaminoethane dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
133.02
Beilstein:
3665235
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

>300 °C (lit.)

SMILES string

Cl[H].Cl[H].NCCN

InChI

1S/C2H8N2.2ClH/c3-1-2-4;;/h1-4H2;2*1H

InChI key

OHHBFEVZJLBKEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinichi Mochizuki et al.
Biochimica et biophysica acta, 1828(2), 412-418 (2012-10-25)
Gene therapy is expected to treat various incurable diseases including viral infections, autoimmune disorders, and cancers. Cationic lipids (CL) have been used as carriers of therapeutic DNAs for gene therapy because they can form a complex with DNA and such
Ana Carolina M S Dias et al.
Biosensors & bioelectronics, 44, 216-221 (2013-02-23)
An immunosensor for the non-structural protein 1 (NS1) of the dengue virus based on carbon nanotube-screen printed electrodes (CNT-SPE) was successfully developed. A homogeneous mixture containing carboxylated carbon nanotubes was dispersed in carbon ink to prepare a screen printed working
Yuxiao Zhang et al.
Nanoscale, 4(24), 7760-7765 (2012-11-10)
Si quantum dots (SiQDs) based nanospheres (SiNSs) were prepared via a novel synthetic strategy. These SiNSs were demonstrated to possess unique dot spacing dependent photoluminescence (PL) up-conversion and surface dependent (N modified surface) down-converted PL. It was demonstrated that a
Kavita Sablok et al.
Journal of hazardous materials, 248-249, 322-328 (2013-02-19)
Binding of electron-deficient trinitrotoluene (TNT) to the electron rich amine groups on a substrate form specific charge-transfer Jackson-Meisenheimer (JM) complex. In the present work, we report formation of specific JM complex on amine functionalized reduced graphene oxide/carbon nanotubes- (a-rGO/CNT) nanocomposite
Nicolò Mauro et al.
Macromolecular bioscience, 13(3), 332-347 (2012-12-15)
This paper reports on the synthesis and physico-chemical, mechanical, and biological characterization of two sets of poly(amidoamine) (PAA) hydrogels with potential as scaffolds for in vivo peripheral nerve regeneration. They are obtained by polyaddition of piperazine with N,N'-methylenebis(acrylamide) or 1,4-bis(acryloyl)piperazine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service