All Photos(2)
About This Item
Linear Formula:
(CH3)2C=NOH
CAS Number:
Molecular Weight:
73.09
Beilstein:
1560146
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
grade
reagent grade
product line
Vetec™
Assay
98%
bp
135 °C (lit.)
mp
60-63 °C (lit.)
density
0.901 g/mL at 25 °C (lit.)
SMILES string
C\C(C)=N/O
InChI
1S/C3H7NO/c1-3(2)4-5/h5H,1-2H3
InChI key
PXAJQJMDEXJWFB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Carc. 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Sens. 1B
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C Kohl et al.
Carcinogenesis, 13(7), 1091-1094 (1992-07-01)
The hepatocarcinogenicity of acetoxime has been tentatively linked with its metabolic oxidation to the potent genotoxicant and carcinogen propane 2-nitronate (P2-N). In order to test the hypothesis that acetoxime is metabolized to P2-N, the oxime (20 mM) was incubated with
Stefanie Zorbas-Seifried et al.
Molecular pharmacology, 71(1), 357-365 (2006-10-20)
The presence of cis-configured exchangeable ligands has long been considered a prerequisite for antitumor activity of platinum complexes, but over the past few years, several examples violating this structure-activity relationship have been recognized. We report here on studies with the
S S Mirvish et al.
Journal of the National Cancer Institute, 69(4), 961-962 (1982-10-01)
Acetoxime was tested for carcinogenicity by chronic administration in the drinking water to male and female outbred MRC-Wistar rats. The dose of 1.0 g/liter was administered 5 days/week for 18 months (total dose, 6.2--7.0 g/rat). The test compound induced benign
M Rysková et al.
Folia biologica, 43(1), 19-24 (1997-01-01)
The genotoxic effects of N-nitroso-N-methylurea (MNU) and acetone oxime (ACOX) were tested in the Somatic Mutation and Recombination Test (SMART) in Drosophila melanogaster. We have performed the same assay on transgenic flies expressing the human gene encoding a glutathione S-transferase
N Guo et al.
Carcinogenesis, 11(9), 1659-1662 (1990-09-01)
The hepatocarcinogens 2-nitropropane and acetoxime have previously been found to induce a specific and qualitatively identical pattern of base damage in rat liver DNA and RNA, including the induction of increased levels of 8-hydroxyguanine. Because both 2-nitropropane and acetoxime are
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service