Skip to Content
Merck
All Photos(1)

Documents

V900579

Sigma-Aldrich

N-Acetyl-L-leucine

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
173.21
Beilstein:
1724849
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

form

crystalline powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

187-190 °C (lit.)

SMILES string

CC(C)C[C@H](NC(C)=O)C(O)=O

InChI

1S/C8H15NO3/c1-5(2)4-7(8(11)12)9-6(3)10/h5,7H,4H2,1-3H3,(H,9,10)(H,11,12)/t7-/m0/s1

InChI key

WXNXCEHXYPACJF-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three-dimensional solid-state NMR experiment that correlates the chemical shift and dipolar coupling frequencies of two heteronuclei.
A Ramamoorthy et al.
Journal of magnetic resonance. Series B, 107(1), 88-90 (1995-04-01)
Silke Dorner et al.
Nucleic acids research, 33(9), 3065-3071 (2005-05-27)
The ribosome is a large RNP complex but its main enzymatic activity, the peptidyl transferase, is a ribozyme. As many RNA enzymes use divalent metal ions in catalysis, one of the hypotheses put forward proposed that metal ions might aid
T Gopinath et al.
Journal of the American Chemical Society, 131(16), 5754-5756 (2009-04-09)
We present a new method for enhancing the sensitivity in static solid-state NMR experiments for a gain in signal-to-noise ratio of up to 40%. This sensitivity enhancement is different from the corresponding solution NMR sensitivity enhancement schemes and is achieved
Chin H Wu et al.
The Journal of chemical physics, 128(5), 052312-052312 (2008-02-13)
The acquisition and analysis of high resolution one- and two-dimensional solid-state nuclear magnetic resonance (NMR) spectra without chemical shift frequencies are described. Many variations of shiftless NMR spectroscopy are feasible. A two-dimensional experiment that correlates the dipole-dipole and dipole-dipole couplings
P Benard et al.
European journal of drug metabolism and pharmacokinetics, 26(1-2), 71-76 (2001-09-14)
Acetyl-DL-leucine [2-14C]: Tanganil a antivertigal drug has been injected intravenously in two macaca monkeys which have been sacrificed 2 and 5 minutes later. Radioactivity distribution has been studied by autoradiography in the brain. An important uptake of radioactivity is localized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service