V900533
Bibenzyl
Vetec™, reagent grade, 98%
Synonym(s):
1,2-Diphenylethane, Dibenzyl
About This Item
Recommended Products
grade
reagent grade
product line
Vetec™
Assay
98%
autoignition temp.
896 °F
bp
284 °C (lit.)
mp
50-53 °C (lit.)
density
1.014 g/mL at 25 °C (lit.)
SMILES string
C(Cc1ccccc1)c2ccccc2
InChI
1S/C14H14/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-10H,11-12H2
InChI key
QWUWMCYKGHVNAV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service