Skip to Content
Merck
All Photos(3)

Key Documents

V900477

Sigma-Aldrich

HEPES

99.5%, crystalline powder, reagent grade, Vetec

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Product V900477 is not currently sold in your country. Contact Technical Service

Product Name

HEPES, Vetec, reagent grade, 99.5%

grade

reagent grade

product line

Vetec

Assay

99.5%

form

crystalline powder

pH

5.0-6.5 (25 °C, 238 g/L)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

water: 500 mg/mL, clear, colorless

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HEPES, a zwitterionic buffer having a pH range of 6.8-8.2, is one among the 20 Good′s buffers. It′s commonly used in cell culture applications as it helps in maintaining the physiological pH.[1][2]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Acid-base buffering in organ preservation solutions as a function of temperature: new parameters for comparing buffer capacity and efficiency
Baicu SC and Taylor MJ
Cryobiology, 45(1), 33-48 (2002)
Evidence-based guidelines for controlling pH in mammalian live-cell culture systems
Michl J, et al.
Communications biology, 2(1), 1-12 (2019)
P Dey et al.
Oncogene, 33(33), 4213-4225 (2013-10-01)
Estrogen receptor β (ERβ) is emerging as a critical factor in understanding prostate cancer biology. Although reduced in prostate cancer above Gleason grade 3, ERβ is a potential drug target at the initial stage of the disease. In human prostate
Bérangère Pinan-Lucarré et al.
Nature, 511(7510), 466-470 (2014-06-05)
Because most neurons receive thousands of synaptic inputs, the neuronal membrane is a mosaic of specialized microdomains where neurotransmitter receptors cluster in register with the corresponding presynaptic neurotransmitter release sites. In many cases the coordinated differentiation of presynaptic and postsynaptic
Frédéric Perros et al.
Journal of the American College of Cardiology, 65(7), 668-680 (2015-02-14)
Endothelial cell (EC) dysfunction plays a central role in the pathogenesis of pulmonary arterial hypertension (PAH), promoting vasoconstriction, smooth muscle proliferation, and inflammation. This study sought to test the hypothesis that nebivolol, a β1-antagonist and β2,3-agonist, may improve PAH and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service