Skip to Content
Merck
All Photos(3)

Key Documents

V900471

Sigma-Aldrich

Adenine

Vetec, reagent grade, ≥99%

Synonym(s):

6-Aminopurine, Vitamin B4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
Beilstein:
5777
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Product V900471 is not currently sold in your country. Contact Technical Service

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

mp

>360 °C (lit.)

SMILES string

Nc1ncnc2[nH]cnc12

InChI

1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)

InChI key

GFFGJBXGBJISGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Didier Wion et al.
Nature reviews. Microbiology, 4(3), 183-192 (2006-02-21)
N(6)-methyl-adenine is found in the genomes of bacteria, archaea, protists and fungi. Most bacterial DNA adenine methyltransferases are part of restriction-modification systems. Certain groups of Proteobacteria also harbour solitary DNA adenine methyltransferases that provide signals for DNA-protein interactions. In gamma-proteobacteria
Henry L Y Chan et al.
Gastroenterology, 146(5), 1240-1248 (2014-01-28)
Little is known about the benefit of antiviral therapy for hepatitis B e antigen (HBeAg)-positive patients with high viral load and normal levels of alanine aminotransferase. We evaluated the effects of single and combination therapies in immune-tolerant patients with chronic
Georg Behrens et al.
AIDS patient care and STDs, 28(4), 168-175 (2014-03-26)
The once daily, single-tablet regimen (STR) combining rilpivirine (RPV), emtricitabine (FTC), and tenofovir disoproxil fumarate (TDF) provides a simplified treatment option for antiretroviral therapy (ART)-naïve patients with baseline HIV-1 RNA (BLVL) of ≤100,000 copies/mL. The aim of this analysis is
Ravindra K Gupta et al.
The Journal of infectious diseases, 207 Suppl 2, S101-S106 (2013-05-25)
Recent data from studies on treatment as prevention (TasP) and preexposure prophylaxis (PrEP) show that antiretroviral drugs can be used in prevention, as well as in treatment. The movement from first-generation antiretroviral therapy (ART) coformulations based on thymidine analogues to
Michinori Matsuo et al.
Journal of molecular and cellular cardiology, 38(6), 907-916 (2005-05-25)
ATP-sensitive potassium (K(ATP)) channels are regulated by adenine nucleotides to convert changes in cellular metabolic levels into membrane excitability. Hence, elucidation of interaction of SUR and Kir6.x with adenine nucleotides is an important issue to understand the molecular mechanisms underlying

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service