Skip to Content
Merck
All Photos(3)

Documents

V900413

Sigma-Aldrich

Hexadecyltrimethylammonium bromide

Vetec, reagent grade, 96%

Synonym(s):

CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(Br)(CH3)3
CAS Number:
Molecular Weight:
364.45
Beilstein:
3598189
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

96%

mol wt

364.45 g/mol

mp

248-251 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

LZZYPRNAOMGNLH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Surface-active agent enhanced FRET effect Cu-doped NH(2)-MIL-88(Fe) for highly sensitive detection of 3-nitro-L-tyrosine.: This study explores the enhancement of Förster Resonance Energy Transfer (FRET) effects in a Cu-doped NH(2)-MIL-88(Fe) framework facilitated by surface-active agents, significantly boosting the sensitivity for detecting 3-nitro-L-tyrosine, a biomarker for nitrosative stress. The presence of hexadecyltrimethylammonium bromide contributes to the increased efficiency and selectivity of the sensor system (Xu et al., 2024).

  • Engineering Interconnected Open-Porous Particles via Microfluidics Using Bijel Droplets as Structural Templates.: This article details the creation of interconnected open-porous particles using bijel droplets as templates in a microfluidic setup. The study highlights the use of hexadecyltrimethylammonium bromide in stabilizing the interface, which is crucial for the structural integrity and functionality of the resulting porous materials (Masaoka et al., 2024).

  • Stabilization of heavy metals in solid waste and sludge pyrolysis by intercalation-exfoliation modified vermiculite.: This research introduces a method for stabilizing heavy metals in pyrolyzed waste using modified vermiculite. Hexadecyltrimethylammonium bromide plays a critical role in the exfoliation and intercalation processes, enhancing the material′s ability to trap and stabilize toxic metals, thereby preventing environmental contamination (Yang et al., 2024).

  • Efficiency of single pharmaceutical surfactants to mimic intestinal biorelevant media solubilization and dissolution of etravirine: Comparison of intrinsic and film dissolution models.: This study evaluates the solubilization and dissolution properties of etravirine in intestinal biorelevant media using different surfactants, including hexadecyltrimethylammonium bromide. The results provide valuable insights into optimizing drug formulations for enhanced bioavailability (Oktay and Polli, 2024).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

471.2 °F - closed cup

Flash Point(C)

244 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A T Nasr et al.
Physics in medicine and biology, 60(12), 4685-4704 (2015-05-29)
In this study, recipe optimization of Leuco Crystal Violet (LCV) micelle gels made with the surfactant Cetyl Trimethyl Ammonium Bromide (CTAB) and the chemical sensitizer 2,2,2-trichloroethanol (TCE) was aided by a two-level three-factor designed experiment. The optimized recipe contains 0.75
Armando Maestro et al.
Langmuir : the ACS journal of surfaces and colloids, 31(23), 6289-6297 (2015-05-15)
We address the rheology of assemblies of surfactant-decorated silica nanoparticles irreversibly adsorbed at the gas/liquid interface. Positively charged surfactant molecules (such as CTAB) bind to silica nanoparticle surfaces, and the resulting particle-surfactant complexes adsorb at gas/liquid interfaces. The surfactant molecules
Fay-Wei Li et al.
Nature communications, 6, 7852-7852 (2015-07-29)
Phytochromes are red/far-red photoreceptors that play essential roles in diverse plant morphogenetic and physiological responses to light. Despite their functional significance, phytochrome diversity and evolution across photosynthetic eukaryotes remain poorly understood. Using newly available transcriptomic and genomic data we show
Moritz Tebbe et al.
Faraday discussions, 181, 243-260 (2015-05-08)
We demonstrate the large-scale organisation of anisotropic nanoparticles into linear assemblies displaying optical anisotropy on macroscopic areas. Monodisperse gold nanorods with a hydrophilic protein shell are arranged by dip-coating on wrinkled surfaces and subsequently transferred to indium tin oxide (ITO)
Carina Hillgruber et al.
The Journal of experimental medicine, 212(8), 1255-1266 (2015-07-15)
Spontaneous organ hemorrhage is the major complication in thrombocytopenia with a potential fatal outcome. However, the exact mechanisms regulating vascular integrity are still unknown. Here, we demonstrate that neutrophils recruited to inflammatory sites are the cellular culprits inducing thrombocytopenic tissue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service