Skip to Content
Merck
All Photos(3)

Key Documents

V900404

Sigma-Aldrich

PIPES

Vetec, reagent grade, ≥99%

Synonym(s):

1,4-Piperazinediethanesulfonic acid, Piperazine-1,4-bis(2-ethanesulfonic acid), Piperazine-N,N′-bis(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O6S2
CAS Number:
Molecular Weight:
302.37
Beilstein:
817713
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Product V900404 is not currently sold in your country. Contact Technical Service

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder or crystals

useful pH range

6.1-7.5

pKa (25 °C)

6.8

mp

>300 °C (lit.)

solubility

1 M NaOH: 0.25 g/mL, clear, colorless

SMILES string

OS(=O)(=O)CCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O6S2/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16/h1-8H2,(H,11,12,13)(H,14,15,16)

InChI key

IHPYMWDTONKSCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronique Adoue et al.
Molecular systems biology, 10, 754-754 (2014-10-19)
Most complex disease-associated genetic variants are located in non-coding regions and are therefore thought to be regulatory in nature. Association mapping of differential allelic expression (AE) is a powerful method to identify SNPs with direct cis-regulatory impact (cis-rSNPs). We used
Leticia A Montoya et al.
Analytical chemistry, 86(12), 6032-6039 (2014-05-24)
Sulfhydryl-containing compounds, including thiols and hydrogen sulfide (H2S), play important but differential roles in biological structure and function. One major challenge in separating the biological roles of thiols and H2S is developing tools to effectively separate the reactivity of these
Nicholas Light et al.
Epigenetics, 9(9), 1238-1251 (2014-07-24)
Allele-specific (AS) assessment of chromatin has the potential to elucidate specific cis-regulatory mechanisms, which are predicted to underlie the majority of the known genetic associations to complex disease. However, development of chromatin landscapes at allelic resolution has been challenging since
J Mano et al.
Biochimica et biophysica acta, 1480(1-2), 374-376 (2000-09-27)
Arabidopsis thaliana P1 protein was crystallized using the hanging drop vapor-diffusion method in 0.1 M piperazine-1, 4-bis(2-ethanesulfonic acid) buffer, containing 14% polyethylene glycol 6000 and 0.2 M magnesium acetate at pH 6.5 and 20 degrees C. The crystals are orthorhombic
V D Prokopieva et al.
Alcohol and alcoholism (Oxford, Oxfordshire), 35(1), 44-48 (2000-02-24)
The effects of carnosine and related compounds on erythrocytes from alcoholics were studied. In their presence, erythrocytes showed an increased ability to resist haemolysis and showed a more normal morphology, with carnosine and N-acetyl-carnosine being the most effective compounds. These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service