Skip to Content
Merck
All Photos(2)

Key Documents

V900372

Sigma-Aldrich

Hygromycin B from Streptomyces hygroscopicus

Vetec, reagent grade

Synonym(s):

Hygrovetine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37N3O13
CAS Number:
Molecular Weight:
527.52
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
Product V900372 is not currently sold in your country. Contact Technical Service

grade

reagent grade

product line

Vetec

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

viruses

storage temp.

2-8°C

SMILES string

CN[C@H]1C[C@@H](N)[C@H](O)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@@H]3O[C@]4(O[C@H]([C@H](N)CO)[C@H](O)[C@H](O)[C@H]4O)O[C@H]23)[C@@H]1O

InChI

1S/C20H37N3O13/c1-23-7-2-5(21)9(26)15(10(7)27)33-19-17-16(11(28)8(4-25)32-19)35-20(36-17)18(31)13(30)12(29)14(34-20)6(22)3-24/h5-19,23-31H,2-4,21-22H2,1H3/t5-,6-,7+,8-,9+,10-,11+,12-,13+,14-,15-,16+,17+,18-,19+,20+/m1/s1

InChI key

GRRNUXAQVGOGFE-XKIAHZFYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • An engineered Streptomyces hygroscopicus aph 7 gene mediates dominant resistance against hygromycin B in Chlamydomonas reinhardtii: This study underscores the genetic engineering applications of Hygromycin B resistance genes, facilitating algae research and potentially biofuel production, illustrating the broad utility of Hygromycin B in genetic engineering and selection protocols (Berthold P et al., 2002).

Biochem/physiol Actions

Mode of Action: The product acts by inhibiting protein synthesis by inducing the misreading of the m-RNA template in the prokaryote, with the potency to inhibit translation.

Antimicrobial Spectrum: Hygromycin B acts against bacteria, fungi and higher eukaryotic cells.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Liu et al.
Plant cell reports, 32(1), 77-87 (2012-11-01)
KEY MESSAGE : Efficient Agrobacterium -mediated genetic transformation for investigation of genetic and molecular mechanisms involved in inflorescence architectures in Cornus species. Cornus canadensis is a subshrub species in Cornus, Cornaceae. It has recently become a favored non-model plant species
Detection of probable marker-free transgene-positive rice plants resistant to rice tungro disease from backcross progenies of transgenic Pusa Basmati 1.
Somnath Roy et al.
Journal of genetics, 91(2), 213-218 (2012-09-04)
Goetz Hensel et al.
BMC plant biology, 12, 171-171 (2012-09-26)
While the genetic transformation of the major cereal crops has become relatively routine, to date only a few reports were published on transgenic triticale, and robust data on T-DNA integration and segregation have not been available in this species. Here
Faisal Asghar Khattak et al.
BMC microbiology, 12, 204-204 (2012-09-13)
The genus Mycobacterium (M.) comprises highly pathogenic bacteria such as M. tuberculosis as well as environmental opportunistic bacteria called non-tuberculous mycobacteria (NTM). While the incidence of tuberculosis is declining in the developed world, infection rates by NTM are increasing. NTM
Bharat P Gurale et al.
The Journal of organic chemistry, 77(13), 5801-5807 (2012-06-06)
Concise and efficient syntheses of the aminocyclitol cores of hygromycin A (HMA) and methoxyhygromycin (MHM) have been achieved starting from readily available myo-inositol. Reductive cleavage of myo-inositol orthoformate to the corresponding 1,3-acetal, stereospecific introduction of the amino group via the

Articles

LC/LC-MS method identifies 8 aminoglycosides in pork using Discovery® DSC-18 SPE and Ascentis® Express C18 UHPLC, per Chinese standards.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service