Skip to Content
Merck
All Photos(3)

Key Documents

V900351

Sigma-Aldrich

5-Fluorocytosine

Vetec, reagent grade, 99%

Synonym(s):

4-amino-5-fluoro-2(1H)-pyrimidinone, Flucytosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4FN3O
CAS Number:
Molecular Weight:
129.09
Beilstein:
127285
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Product V900351 is not currently sold in your country. Contact Technical Service

grade

reagent grade

product line

Vetec

Assay

99%

mp

298-300 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

NC1=NC(=O)NC=C1F

InChI

1S/C4H4FN3O/c5-2-1-7-4(9)8-3(2)6/h1H,(H3,6,7,8,9)

InChI key

XRECTZIEBJDKEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nucleoside analog that has antifungal activities. 5-FC is deaminated by cytosine deaminase to product 5-fluorouracil, resulting in RNA miscoding. 5-Fluorocytosine inhibits DNA and RNA synthesis and interferes with ribosomal protein synthesis.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna K Kaufmann et al.
The Journal of investigative dermatology, 133(4), 1034-1042 (2012-12-12)
Effective treatment modalities for advanced melanoma are desperately needed. An innovative approach is virotherapy, in which viruses are engineered to infect cancer cells, resulting in tumor cell lysis and an amplification effect by viral replication and spread. Ideally, tumor selectivity
Francesco Imperi et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(18), 7458-7463 (2013-04-10)
Although antibiotic resistance represents a public health emergency, the pipeline of new antibiotics is running dry. Repurposing of old drugs for new clinical applications is an attractive strategy for drug development. We used the bacterial pathogen Pseudomonas aeruginosa as a
Radha Rajasingham et al.
PLoS medicine, 9(9), e1001316-e1001316 (2012-10-12)
Cryptococcal meningitis (CM) is the most common form of meningitis in Africa. World Health Organization guidelines recommend 14-d amphotericin-based induction therapy; however, this is impractical for many resource-limited settings due to cost and intensive monitoring needs. A cost-effectiveness analysis was
Cestmir Altaner et al.
International journal of cancer, 134(6), 1458-1465 (2013-09-17)
Suicide gene therapy mediated by mesenchymal stem cells with their ability to engraft into tumors makes these therapeutic stem cells an attractive tool to activate prodrugs directly within the tumor mass. In this study, we evaluated the therapeutic efficacy of
C Wang et al.
Cancer gene therapy, 19(11), 796-801 (2012-09-29)
In our previous works, we demonstrated that human neural stem cells (NSCs) transduced with the cytosine deaminase (CD) gene showed remarkable 'bystander killer effect' on glioma and medulloblastoma cells after administration of the prodrug 5-fluorocytosine (5-FC). In addition, herpes simplex

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service