Skip to Content
Merck
All Photos(3)

Key Documents

V900312

Sigma-Aldrich

Trizma® hydrochloride

Vetec, reagent grade, ≥99%

Synonym(s):

TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥99%

form

crystalline powder

useful pH range

7.0-9.0

pKa (25 °C)

8.1

solubility

water: 0.666 g/mL, clear, colorless

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany
Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yushuai Wang et al.
iScience, 24(10), 103148-103148 (2021-10-15)
Tropical plants have adapted to strong solar ultraviolet (UV) radiation. Here we compare molecular responses of two tropical mangroves Avecennia marina and Rhizophora apiculata to high-dose UV-B. Whole-genome bisulfate sequencing indicates that high UV-B induced comparable hyper- or hypo-methylation in
Weixiao Yin et al.
Bio-protocol, 8(9), e2839-e2839 (2018-05-05)
Here, we describe a method that can be used to evaluate the function of predicted signal peptides. This method utilizes the yeast Saccharomyces cerevisiae YTK12 strain and pSUC2 vector in which the pSUC2 vector with fused predicted signal peptide is
Chao Zhang et al.
Bio-protocol, 8(20), e3060-e3060 (2018-10-20)
Virus inoculation is a basic experimental procedure to evaluate the resistance of a rice variety or a transgenic material upon virus infection. We recently demonstrated that Rice ragged stunt virus (RRSV), an oryzavirus that is transmitted by brown planthopper (BPH)
Lijia Zheng et al.
Bio-protocol, 8(9), e2840-e2840 (2018-05-05)
MicroRNAs (miRNAs) play important roles in plant growth, development, and response to infection by microbes. Double-stranded RNA binding protein 1 (DRB1) facilitates the processing of primary miRNA transcripts into mature miRNAs. Recently, we found that NS3 protein encoded by rice
Tibor Maliar et al.
Chemistry & biodiversity, 12(11), 1678-1695 (2015-11-17)
The content of biogenic compounds and the biological activities of barley (Hordeum vulgare L.)-grain extracts was evaluated. The sufficiently large and heterogeneous set of barley genotypes (100 accessions) enabled the selection of special genotypes interesting for potential industrial, pharmaceutical, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service