Skip to Content
Merck
All Photos(3)

Documents

V900303

Sigma-Aldrich

L-Arginine monohydrochloride

≥98%, Vetec

Synonym(s):

(S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
Beilstein:
3631658
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

product name

L-Arginine monohydrochloride, Vetec, reagent grade, ≥98%

grade

reagent grade

product line

Vetec

Assay

≥98%

form

powder

color

white

mp

226-230 °C

solubility

water: 50 mg/mL, clear, colorless

density

1.42 g/cm3 at 20 °C

SMILES string

Cl[H].N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1

InChI key

KWTQSFXGGICVPE-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Product-Specific Impact of Viscosity Modulating Formulation Excipients During Ultra-High Concentration Biotherapeutics Drug Product Development.: This paper highlights the role of ʟ-Arginine monohydrochloride in modulating viscosity in biotherapeutics, crucial for the development of high-concentration formulations (Rodrigues et al., 2021).

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Di Somma et al.
Endocrine, 49(2), 492-502 (2014-11-10)
The aim of the study is to clarify the relationship between adipose tissue dysfunction, metabolic profile and growth hormone (GH)/insulin-like growth factor (IGF)-I secretion in healthy adult subjects. We investigated the metabolic profile in a cohort of 231 consecutive healthy
C B Christiansen et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 47(7), 537-543 (2015-01-21)
TLQP-21 is a VGF-derived neuropeptide proposed to be involved in regulation of metabolism. More specifically it has been suggested that TLQP-21 has the ability to enhance glucose stimulated insulin secretion, making it a candidate for treatment of patients with type
Jenna Gallie et al.
PLoS biology, 13(3), e1002109-e1002109 (2015-03-13)
Phenotype switching is commonly observed in nature. This prevalence has allowed the elucidation of a number of underlying molecular mechanisms. However, little is known about how phenotypic switches arise and function in their early evolutionary stages. The first opportunity to
N E Paul et al.
Bio-medical materials and engineering, 25(3), 289-297 (2015-09-26)
Adipose tissue engineering is a promising solution for the reconstruction of soft tissue defects. An insufficient neovascularisation within the scaffolds that leads to necrosis and tissue loss is still a major shortcoming of current tissue engineering attempts. Biomaterials, which release
Nivaldo Ribeiro Villela et al.
Journal of translational medicine, 12, 232-232 (2014-08-26)
Relative hypovolemia is frequently found in early stages of severe sepsis and septic shock and prompt and aggressive fluid therapy has become standard of care improving tissue perfusion and patient outcome. This paper investigates the role of the nitric oxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service