Skip to Content
Merck
All Photos(1)

Key Documents

V900236

Sigma-Aldrich

Triethylene glycol

Vetec, reagent grade, 98%

Synonym(s):

Triglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)2CH2CH2OH
CAS Number:
Molecular Weight:
150.17
Beilstein:
969357
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
reagent grade
bp:
125-127 °C/0.1 mmHg (lit.)
vapor pressure:
<0.01 mmHg ( 20 °C)

grade

reagent grade

vapor density

5.2 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

product line

Vetec

Assay

98%

form

viscous liquid

autoignition temp.

699 °F

expl. lim.

9.2 %

dilution

(for analytical testing)

refractive index

n20/D 1.455 (lit.)

bp

125-127 °C/0.1 mmHg (lit.)

mp

−7 °C (lit.)

density

1.124 g/mL at 20 °C (lit.)

SMILES string

OCCOCCOCCO

InChI

1S/C6H14O4/c7-1-3-9-5-6-10-4-2-8/h7-8H,1-6H2

InChI key

ZIBGPFATKBEMQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

330.8 °F - closed cup

Flash Point(C)

166 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kengo Arai et al.
Molecules (Basel, Switzerland), 25(20) (2020-10-21)
Hydroxyethyl cellulose (HeC) maintains high water solubility over a wide temperature range even in a high temperature region where other nonionic chemically modified cellulose ethers, such as methyl cellulose (MC) and hydroxypropylmethyl cellulose (HpMC), demonstrate cloud points. In order to
Jianji Wang et al.
The journal of physical chemistry. B, 115(17), 4955-4962 (2011-04-13)
Material preparation in ionic liquids and environmental pollution control by ionic liquids are often closely dependent on the aggregation behavior of ionic liquids in solution. In the present work, conductivity, fluorescence probe, and dynamic light scattering techniques have been used
Amit Vaish et al.
Small (Weinheim an der Bergstrasse, Germany), 7(10), 1471-1479 (2011-05-04)
Soft-lithography-based techniques are widely used to fabricate microarrays. Here, the use of microcontact insertion printing is described, a soft-lithography method specifically developed for patterning at the dilute scales necessary for highly selective biorecognition. By carefully tuning the polar surface energy
Babak Karimi et al.
Chemical communications (Cambridge, England), 47(27), 7686-7688 (2011-06-11)
A novel water-soluble NHC-Pd polymer with triethylene glycol legs was developed as a water soluble and highly recyclable catalyst that shows high catalytic activity in the Suzuki-Miyaura coupling of aryl chlorides in high yields in water at room temperature.
Lei Dong et al.
Nanoscale, 3(9), 3447-3461 (2011-07-26)
A p/n heterojunction is the basic setup for light-electric conversion. It has been widely accepted that the ideal configuration for organic photovoltaics is formed by the joint of a pair of long-range continuous but nanometre-wide phases consisting of electron-donating (D)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service