Skip to Content
Merck
All Photos(2)

Key Documents

V900198

Sigma-Aldrich

Magnesium nitrate hexahydrate

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
256.41
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

reagent grade

vapor density

6 (vs air)

product line

Vetec

Assay

98%

form

solid

mp

89 °C (dec.) (lit.)

SMILES string

O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Mg.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

MFUVDXOKPBAHMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed Abdellatif Bensegueni et al.
Acta crystallographica. Section C, Structural chemistry, 71(Pt 3), 222-228 (2015-03-04)
Two alkaline earth-tetrazole compounds, namely catena-poly[[[triaquamagnesium(II)]-μ-5,5'-(azanediyl)ditetrazolato-κ(3)N(1),N(1'):N(5)] hemi{bis[μ-5,5'-(azanediyl)ditetrazolato-κ(3)N(1),N(1'):N(2)]bis[triaquamagnesium(II)]} monohydrate], {[Mg(C(2)HN(9))(H(2)O)(3)][Mg(2)(C(2)HN(9))(2)(H(2)O)(6)]0.5·H(2)O}n, (I), and bis[5-(pyrazin-2-yl)tetrazolate] hexaaquamagnesium(II), (C(5)H(3)N(6))[Mg(H(2)O)(6)], (II), have been prepared under hydrothermal conditions. Compound (I) is a mixed dimer-polymer based on magnesium ion centres and can be regarded as the first example
Sheng Tang et al.
Journal of chromatography. A, 1373, 31-39 (2014-12-02)
Layered double oxide hollow spheres (LDO-HSs) were synthesized and employed as a dissolvable sorbent in dispersive solid-phase extraction (DSPE) to extract eleven United States Environmental Protection Agency's priority phenols from aqueous samples. With their higher specific surface area, LDO-HSs showed
Zhaoying Wu et al.
Colloids and surfaces. B, Biointerfaces, 120, 38-46 (2014-06-07)
Mesoporous magnesium silicate (m-MS) was synthesized, and the in vitro degradability, bioactivity and primary cell responses to m-MS were investigated. The results suggested that the m-MS with mesoporous channels of approximately 5nm possessed the high specific surface area of 451.0m(2)/g
Raquel C B Marques et al.
Talanta, 129, 594-599 (2014-08-17)
Human epidermal growth factor receptor 2 (HER2) is a breast cancer biomarker that plays a major role in promoting breast cancer cell proliferation and malignant growth. The extracellular domain (ECD) of HER2 can be shed into the blood stream and
M Fátima Pina et al.
International journal of pharmaceutics, 481(1-2), 114-124 (2015-01-17)
In this study, we evaluate the use of theoretical thermodynamic analysis of amorphous paroxetine hydrochloride (HCl) as well as experimental assessment in order to identify the most promising approach to stability and dissolution behaviour prediction, particularly in relation to stoichiometric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service