V900009
Polyvinylpyrrolidone
Vetec™, reagent grade, average mol wt 10,000
Synonym(s):
PVP, Polyvidone, Povidone
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
grade
reagent grade
product line
Vetec™
mol wt
average mol wt 10,000
SMILES string
C=CN1CCCC1=O
InChI
1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2
InChI key
WHNWPMSKXPGLAX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chun-Mei Zhao et al.
Environmental toxicology and chemistry, 32(4), 913-919 (2013-01-25)
The toxicity of manufactured silver nanoparticles (AgNPs) has been widely studied, but the influence of AgNPs on the major ions (such as sodium [Na] and calcium [Ca]) regulations are unknown. In the present study, a freshwater cladoceran Daphnia magna was
Liyun Ren et al.
Nanoscale, 5(6), 2337-2345 (2013-02-09)
PLLA fibrous tissue scaffolds with controlled fiber nanoscale surface roughness are fabricated with a novel centrifugal jet spinning process. The centrifugal jet spinning technique is a highly efficient synthesis method for micron- to nano-sized fibers with a production rate up
Doris Bach et al.
Journal of photochemistry and photobiology. B, Biology, 120, 74-81 (2013-03-08)
Photodynamic therapy (PDT) is a local tumour treatment accepted for a number of indications. PDT operates via the cellular stress response through the production of reactive oxygen species and subsequent cellular damage, resulting in cell death. Although PDT-induced signalling and
Liandong Hu et al.
Pakistan journal of pharmaceutical sciences, 26(3), 629-636 (2013-04-30)
Solid dispersion technique has been developed many years for improving solubility of water-insoluble drugs, aiming to achieve a better oral bioavailability. However, this technique exhibits many inconveniences when used for large-scale tableting procedures. The objective of current research work was
Bin Li et al.
Carbohydrate polymers, 92(2), 2033-2040 (2013-02-13)
Amorphous solid dispersions (ASD) of quercetin (Que) in cellulose derivative matrices, carboxymethylcellulose acetate butyrate (CMCAB), hydroxypropylmethylcellulose acetate succinate (HPMCAS), and cellulose acetate adipate propionate (CAAdP) were prepared with the goal of identifying an ASD that effectively increased Que aqueous solution
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service