Skip to Content
Merck
All Photos(1)

Key Documents

V800414

Sigma-Aldrich

L-(+)-Tartaric acid

AR, ≥99%

Synonym(s):

(2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725147
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Product V800414 is not currently sold in your country. Contact Technical Service

grade

AR

vapor density

5.18 (vs air)

product line

Vetec

Assay

≥99%

autoignition temp.

797 °F

mp

170-172 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaharu Sugiura et al.
Organic letters, 16(19), 5172-5175 (2014-09-24)
Enantioselective conjugate addition of styrylboronic acid to dienones was effectively catalyzed by an O-monoacyltartaric acid to afford monostyrylated products with good enantioselectivity. The RCM of the monostyrylated products using the Hoveyda-Grubbs II catalyst afforded optically active cyclopentenones, including a synthetic
Preparation of organotrifluoroborate salts: precipitation-driven equilibrium under non-etching conditions.
Alastair J J Lennox et al.
Angewandte Chemie (International ed. in English), 51(37), 9385-9388 (2012-08-21)
Yanfang Feng et al.
Bioresource technology, 125, 138-144 (2012-10-03)
The aim of this study was to develop a promising and competitive bioadsorbent with the abundant of source, low price and environmentally friendly characters to remove cationic dye from wastewater. The swede rape straw (Brassica napus L.) modified by tartaric
Andrea Bencini et al.
Chemical communications (Cambridge, England), 48(84), 10428-10430 (2012-09-18)
A chiral ditopic polyammonium receptor featuring two [9]aneN(3) moieties separated by a (S)-BINOL linker is able to selectively bind and sense in water (S,S)-tartaric acid over its (R,R)/meso forms.
Anne Toboldt et al.
Applied and environmental microbiology, 78(20), 7347-7357 (2012-08-14)
In this study, the population structure, incidence, and potential sources of human infection caused by the d-tartrate-fermenting variant of Salmonella enterica serovar Paratyphi B [S. Paratyphi B (dT+)] was investigated. In Germany, the serovar is frequently isolated from broilers. Therefore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service